40-50023
xMAP® Sheath Concentrate PLUS, RUO
Synonym(s):
40-50000, 4050000, FLEXMAP 3D, FlexMAP, Luminex, SHEATHFLUID PLUS Concentrate, flexmap, lx100, lx200, sheath fluid, sheathfluid PLUS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
packaging
pkg of 1 L
manufacturer/tradename
Milliplex®
technique(s)
multiplexing: suitable
storage temp.
room temp
General description
The FLEXMAP 3D®, xMAP® INTELLIFLEX RU0 and INTELLIFLEX-DRSE, and Luminex 200™ instruments require sheath fluid PLUS to serve as the delivery medium to transport the sample to the instrument′s optics.
Application
N/A
Legal Information
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
FLEXMAP 3D is a registered trademark of Luminex Corp
Luminex 200 is a trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service