Skip to Content
Merck
All Photos(1)

Documents

324726

Sigma-Aldrich

Endoglycosidase F2, Elizabethkingia meningosepticum, Recombinant, E. coli

Endoglycosidase F2, Elizabethkingia meningosepticum, Recombinant, E. coli cleaves asparagine-linked or free oligomannose and biantennary complex oligosaccharides. It is not active above pH 6.0.

Synonym(s):

Endo-β-N-acetylglucosaminidase F2, Endo F2

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

conjugate

(N-linked)

form

liquid

specific activity

≥20 units/mg protein
≥5 units/mL

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

N-acetylglucosaminidase, α- and β-galactosidase, α-mannosidase, neuraminidases, proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Note: 1 mU = 1 milliunit.
Recombinant, Elizabethkingia meningosepticum endoglycosidase F2 expressed in E. coli. Cleaves asparagine-linked or free oligomannose and biantennary complex oligosaccharides from glycoproteins. It cleaves between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide generating a truncated sugar molecule with one N-acetylglucosamine residue remaining on the asparagine. Less sensitive to protein conformation than N-Glycosidase F (Cat. No. 362185) and therefore is more suitable for deglycosylation of native proteins. This enzyme is not active above pH 6.0.
Recombinant, Elizabethkingia meningosepticum endoglycosidase F2 expressed in E. coli. Cleaves asparagine-linked or free oligomannose and biantennary complex oligosaccharides from glycoproteins. It cleaves between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide generating a truncated sugar molecule with one N-acetylglucosamine residue remaining on the asparagine. Less sensitive to protein conformation than N-Glycosidase F (Cat. No. 362185) and therefore, is more suitable for deglycosylation of native proteins.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will release N-linked oligosaccharides from 1 µmol porcine fibrinogen per min at 37°C, pH 4.5.

Physical form

In 25 mM NaCl, 10 mM sodium acetate buffer, pH 4.5.

Other Notes

Reddy, A., et al. 1998. Glycobiology 8, 633.
Tarentino, A.L., and Plummer, T.H. 1994. Methods Enzymol. 230, 44.
Tarentino, A.L., et al. 1993. J. Biol. Chem. 268, 9702.
Trimble, R.B., and Tarentino, A.L. 1991. J. Biol. Chem. 266, 1646.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Reddy et al.
Glycobiology, 8(6), 633-636 (1998-06-11)
The Endo F2gene was overexpressed in E.coli as a fusion protein joined to the maltose-binding protein. MBP-Endo F2was found in a highly enriched state as insoluble, inactive inclusion bodies. Extraction of the inclusion bodies with 20% acetic acid followed by
R B Trimble et al.
The Journal of biological chemistry, 266(3), 1646-1651 (1991-01-25)
Flavobacterium meningosepticum endo-beta-acetyl-glucosaminidase F preparations have been resolved by hydrophobic interaction chromatography on TSK-butyl resin into at least three activities designated endo F1, endo F2 and endo F3 each with a unique substrate specificity. The 32-kDa endo F1 protein is
A L Tarentino et al.
The Journal of biological chemistry, 268(13), 9702-9708 (1993-05-05)
The genes for Flavobacterium meningosepticum Endo (endoglycosidase) F2 and Endo F3 were cloned, and their nucleotide sequences were determined. The deduced amino acid sequences were verified independently to a large extent by direct peptide microsequencing of 66 and 84% of
Enzymatic deglycosylation of asparagine-linked glycans: purification, properties, and specificity of oligosaccharide-cleaving enzymes from Flavobacterium meningosepticum.
A L Tarentino et al.
Methods in enzymology, 230, 44-57 (1994-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service