218783
Caspase-1, Human, Recombinant, E. coli
Caspase-1, Human, Recombinant, is expressed in E. coli and is fused at the N-terminus to a His•Tag sequence.
Synonym(s):
IL-1β Converting Enzyme, ICE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.77
Recommended Products
recombinant
expressed in E. coli
Quality Level
Assay
≥90% (SDS-PAGE)
form
liquid
specific activity
≥25,000 units/mg protein
≥50,000 units/mL
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
dry ice
storage temp.
−70°C
General description
Research area: APOPTOSIS
Caspases are enzymes that belong to the cysteine protease family. In mammals, researchers have discovered 13 caspases. They enhance chemical reactions by targeting specific substances, guided by aspartic acid. These caspases can be further classified into two groups: apoptotic caspases (caspase-2, 3, 6, 7, 8, 9, and 10) and inflammatory caspases (caspase-1, 4, 5, 11, and 12). Apart from inflammatory organs such as the spleen, lymph nodes, and thymus, caspase-1 is also expressed in adipose tissue, liver, and intestine. Recombinant, human caspase-1 fused at the N-terminus to a His•Tag sequence and expressed in E. coli. Useful for the study of enzyme regulation, cleavage of target substrates, and inhibitor screening. M.W. 10000 and 20000.
Caspases are enzymes that belong to the cysteine protease family. In mammals, researchers have discovered 13 caspases. They enhance chemical reactions by targeting specific substances, guided by aspartic acid. These caspases can be further classified into two groups: apoptotic caspases (caspase-2, 3, 6, 7, 8, 9, and 10) and inflammatory caspases (caspase-1, 4, 5, 11, and 12). Apart from inflammatory organs such as the spleen, lymph nodes, and thymus, caspase-1 is also expressed in adipose tissue, liver, and intestine. Recombinant, human caspase-1 fused at the N-terminus to a His•Tag sequence and expressed in E. coli. Useful for the study of enzyme regulation, cleavage of target substrates, and inhibitor screening. M.W. 10000 and 20000.
Application
Caspase-1 enzyme has been used in mediating the proteolysis of pro-IL-1β for regulating the availability and activation of IL-1β.
Biochem/physiol Actions
Caspases belong to the cysteine proteases family of enzymes that are responsible for regulating apoptosis and pyroptosis, a programmed necrosis. The activation of caspase-1 in inflammasomes initiates an inflammatory response for fighting infections or cellular damage. Furthermore, caspase-1 activated interleukin-1 beta (IL-1β) and interleukin-18 (IL-18) are related to various metabolic disorders such as obesity, atherosclerosis, impairment of insulin action and/or secretion, cancer, and nonalcoholic fatty liver disease. Caspase-1 deficiency could play a role in promoting the growth of tumor cells in colon, ovarian, and prostate cancers.
Warning
Toxicity: Standard Handling (A)
Unit Definition
One unit is defined as the amount of enzyme that will cleave 1.0 pmol Ac-YVAD-pNA (Cat. No. 400025) per min at 30°C, pH 7.4.
Physical form
In 100 mM NaCl, 50 mM HEPES, 10 mM DTT, 1 mM EDTA, 10% glycerol, 0.5% CHAPS, pH 7.4.
Reconstitution
Following initial thaw, aliquot and freeze (-70°C).
Other Notes
Thornberry, N.A. 1994. Methods Enzymol. 244, 615.
Thornberry, N.A., et al. 1992. Nature 356, 768.
Thornberry, N.A., et al. 1992. Nature 356, 768.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Role of Caspase-1 in the Pathogenesis of Inflammatory-Associated Chronic Noncommunicable Diseases
Molla MD, et al.
Journal of Inflammation Research, 13, 749-764 (2020)
Acid-dependent Interleukin-1 (IL-1) Cleavage Limits Available Pro-IL-1? for Caspase-1 Cleavage
Edye MD, et al.
The Journal of Biological Chemistry, 290(42), 25374-25381 (2015)
Caspase-1 as a regulatory molecule of lipid metabolism
Molla MD, et al.
Lipids in Health and Disease, 19(1), 34-34 (2020)
Caspase-1 initiates apoptosis in the absence of gasdermin D
Tsuchiya K, et al.
Nature Communications, 10(1), 2091-2091 (2019)
Pingping Ren et al.
Journal of the American Heart Association, 9(7), e014044-e014044 (2020-04-01)
Background Aortic aneurysms and dissections are highly lethal diseases for which an effective treatment strategy is critically needed to prevent disease progression. The nucleotide-binding oligomerization domain-like receptor pyrin domain containing 3 (NLRP3)-caspase-1 inflammasome cascade was recently shown to play an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service