Skip to Content
Merck
All Photos(2)

Key Documents

1.88050

Supelco

Water Standard

0.01%, Reference Material for coulometric Karl Fischer Titration 1 g ≙ 0.1 mg H₂O Aquastar®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601
EC Index Number:
215-535-7
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Water Standard 0.01 %, Reference Material for coulometric Karl Fischer Titration 1 g ≙ 0.1 mg H₂O Aquastar®

grade

reference material

Quality Level

Agency

ISO 17025

vapor pressure

10 hPa ( 20 °C)

product line

Aquastar®

form

liquid

autoignition temp.

465 °C

expl. lim.

1.7-7.5 % (v/v)

packaging

ampule of 10 × 8 mL

technique(s)

Karl-Fischer titration: suitable
titration: suitable

bp

137 °C/1013 hPa

mp

-34 °C (xylene)

transition temp

flash point 25 °C

solubility

0.2 g/L

density

0.86 g/mL at 20 °C

format

single component solution

storage temp.

15-25°C

General description

The 0.01% water standard is gravimetrically prepared from high-purity water and reagents. Its characterization is performed at an accredited quality control (QC) laboratory according to DIN EN ISO/IEC 17025. The water content of the standard is measured by direct coulometric Karl Fischer titration following ISO 760.

Application

It is used as a reference material to verify the accuracy of coulometric Karl Fischer titrators according to ISO 9001 chapter 7.1.5 “Monitoring and measuring resources”. It can also be used to check the accuracy of measured results according to European Pharmacopeia (Ph.Eur.) chapter 2.5.32 “Water Micro Determination”.

Features and Benefits

  • Packed in convenient 8 mL glass ampules. Each box contains 10 single-use ampules
  • Supplied with a batch-specific certificate of analysis (CoA) having data on exact measured water content of the batch, uncertainty, method of analysis, the NIST batch, and minimum shelf life
  • Measured and qualified in an ISO/IEC 17025 accredited laboratory
  • Directly traceable to SI unit (kg) and verified by secondary reference standards 2890 from NIST
  • Shelf life of 18 months

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals.

Analysis Note

Assay (H₂O): 0.090 - 0.110 mg/g

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. bonjour, avons nous l'information sur la densité du produits en g/cm3? sur la boite ou sur le CoA? merci ref 1.88050

    1 answer
    1. As stated in the Properties section of the product page, the material has a density of 0.86 g/mL at 20°C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service