1.88009
Methanol
CombiMethanol, Solvent for volumetric Karl Fischer titration with one component reagents max. 0.01% H₂O Aquastar®
Synonym(s):
Methanol, MeOH, Hydroxymethane, Methyl alcohol, Carbinol, Methyl alcohol
About This Item
Recommended Products
Product Name
CombiMethanol, Solvent for volumetric Karl Fischer titration with one component reagents max. 0.01% H₂O Aquastar®
vapor density
1.11 (vs air)
Quality Level
vapor pressure
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
product line
Aquastar®
form
liquid
autoignition temp.
725 °F
potency
5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)
expl. lim.
36 %
reaction suitability
reaction type: volumetric
technique(s)
Karl-Fischer titration: suitable
titration: suitable
refractive index
n20/D 1.329 (lit.)
kinematic viscosity
0.54-0.59 cSt(20 °C)
bp
64.7 °C (lit.)
mp
−98 °C (lit.)
transition temp
flash point 9.7 °C
density
0.791 g/mL at 25 °C (lit.)
format
single component solution
storage temp.
15-25°C
SMILES string
CO
InChI
1S/CH4O/c1-2/h2H,1H3
InChI key
OKKJLVBELUTLKV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Analysis Note
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service