1.88011
Titrant 2
1 ml ≙ ca. 2 mg H₂O, titrant for Karl Fischer titration with two component reagents, AQUASTAR®®
Synonym(s):
Titration Solution 2
About This Item
Recommended Products
product name
Titrant 2, titrant for volumetric Karl Fischer titration with two component reagents 1 ml ≙ ca. 2 mg H₂O Aquastar®
Quality Level
product line
Aquastar®
form
liquid
reaction suitability
reaction type: volumetric
concentration
2 mg/mL
technique(s)
Karl-Fischer titration: suitable
titration: suitable
transition temp
flash point <21 °C
density
0.82 g/cm3 at 20 °C
format
two component solution
storage temp.
15-25°C
Related Categories
Packaging
Analysis Note
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT RE 2 Oral - STOT SE 1
Target Organs
Thyroid
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
69.8 °F
Flash Point(C)
< 21 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service