Skip to Content
Merck
All Photos(1)

Key Documents

16-193

Sigma-Aldrich

Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, biotin conjugate

clone JBW301, Upstate®, from mouse

Synonym(s):

H2AXS139P, Histone H2A.X (phospho S139), H2A histone family, member X, H2AX histone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

JBW301, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer139)

Gene Information

human ... H2AX(3014)

General description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures.

Specificity

A broad species cross-reactivity is expected based on conservation of sequence homology.
Recognizes Histone H2A.X phosphorylated at Ser139, Mr 15kDa.

Immunogen

Epitope: Ser139
KLH-conjugated, synthetic peptide (CKATQA[pS]QEY) corresponding to amino acids 134-142 of human histone H2A.X. The immunizing sequence has 8 identical amino acids in yeast and mouse.

Application

Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, biotin conjugate is a Mouse Monoclonal Antibody for detection of phospho-Histone H2A.X (Ser139) also known as H2AXS139P, H2AX histone & has been validated in ICC & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated in acid extracted histone from Jurkat cell lysates treated with 0.5mM staurosporine (19-123).

Western Blot Analysis:
0.5-1 μg/mL of this lot detected phosphorylated histone H2A.X (Ser139) in acid extracted histone from Jurkat cell lysates treated with 0.5μM staurosporine (Catalog # 19-123).

Target description

15 kDa

Physical form

Protein G Purified
Purified biotin conjugated mouse monoclonal IgG1 in buffer containing 70% storage buffer (0.02 M Phosphate buffer, pH7.6, 0.25 M NaCl, 0.1% sodium azide) and 30% glycerol

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerolcontaining solutions to become frozen during storage.

Analysis Note

Control
Staurosporine-treated Jurkat cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey M Cloutier et al.
PLoS genetics, 11(10), e1005462-e1005462 (2015-10-29)
Chromosome abnormalities are common in the human population, causing germ cell loss at meiotic prophase I and infertility. The mechanisms driving this loss are unknown, but persistent meiotic DNA damage and asynapsis may be triggers. Here we investigate the contribution
Sascha Schäuble et al.
PloS one, 7(8), e42150-e42150 (2012-08-11)
Primary human fibroblasts in tissue culture undergo a limited number of cell divisions before entering a non-replicative "senescent" state. At early population doublings (PD), fibroblasts are proliferation-competent displaying exponential growth. During further cell passaging, an increasing number of cells become
Tristan Winters et al.
The EMBO journal, 33(11), 1256-1270 (2014-05-07)
The cohesin complex is essential for mitosis and meiosis. The specific meiotic roles of individual cohesin proteins are incompletely understood. We report in vivo functions of the only meiosis-specific STAG component of cohesin, STAG3. Newly generated STAG3-deficient mice of both
Jinting Yan et al.
Journal of cell science, 129(3), 492-501 (2015-12-15)
Human Huntingtin (HTT), a Huntington's disease gene, is highly expressed in the mammalian brain and testis. Simultaneous knockout of mouse Huntingtin (Htt) in brain and testis impairs male fertility, providing evidence for a link between Htt and spermatogenesis; however, the
Maria Delgado-Esteban et al.
Nature communications, 4, 2879-2879 (2013-12-05)
The morphology of the adult brain is the result of a delicate balance between neural progenitor proliferation and the initiation of neurogenesis in the embryonic period. Here we assessed whether the anaphase-promoting complex/cyclosome (APC/C) cofactor, Cdh1--which regulates mitosis exit and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service