14-602-M
PI3 Kinase (p110α/p85α) Protein, active, 20 µg
Active complex of N-terminal His6-tagged recombinant full-length human p110α & untagged, recombinant, full length, human p85α. For use in Kinase Assays.
Synonym(s):
Active PI3 Kinase Protein, PI3 Kinase p110α/p85α
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.26
Recommended Products
biological source
human
Quality Level
recombinant
expressed in baculovirus infected Sf21 cells
mol wt
Mw 125.3 kDa (p110α)
Mw 83.7 kDa (p85α)
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
Gene Information
human ... PIK3CA(5290)
General description
Product Source: Coexpressed by baculovirus in Sf21 insect cells
Research area: Cell signaling
Protein Target: PI3Kα/p85α Target Sub-Family: PI3K/PI4K
Complex of N-terminal His6-tagged recombinant full-length human p110α and untagged, recombinant, full length, human p85α
Protein Target: PI3Kα/p85α Target Sub-Family: PI3K/PI4K
Complex of N-terminal His6-tagged recombinant full-length human p110α and untagged, recombinant, full length, human p85α
Application
PI3 Kinase (p110α/p85α) Protein has been used in in vitro kinase assay.
Biochem/physiol Actions
Phosphoinositide 3-kinases (PI3Ks) are enzymes that play a crucial role in cell signaling. They are responsible for activating signaling pathways that regulate important cellular processes such as cell survival, proliferation, protein synthesis, cell metabolism, cytoskeletal rearrangements, and vesicle trafficking. PI3Ks exhibits both lipid kinase activity (phosphorylation of inositol ring in phosphatidylinositol (PtdIns) lipids) and protein kinase activity (including autophosphorylation).
Packaging
Also available in 250μg size as catalog # 14-602M - Call for pricing and availability.
Quality
routinely evaluated by PIProfiler
Physical form
Ni2+/NTA-agarose
Storage and Stability
6 months at -70°C
Other Notes
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kinetic and structural analyses reveal residues in phosphoinositide 3-kinase ? that are critical for catalysis and substrate recognition
Maheshwari S, et al.
The Journal of biological chemistry, 292(33), 13541-13550 (2017)
PKCalpha reduces the lipid kinase activity of the p110alpha/p85alpha PI3K through the phosphorylation of the catalytic subunit
Sipeki S, et al.
Biochemical and Biophysical Research Communications, 339(1), 122-125 (2006)
Christina M Buchanan et al.
PloS one, 8(8), e71337-e71337 (2013-08-13)
In addition to lipid kinase activity, the class-I PI 3-kinases also function as protein kinases targeting regulatory autophosphorylation sites and exogenous substrates. The latter include a recently identified regulatory phosphorylation of the GM-CSF/IL-3 βc receptor contributing to survival of acute
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service