1.40197
EcoCult® Half FRASER broth with antibiotics
ISO 11290, APHA, suitable for microbiology, pathogen testing
Synonym(s):
Demi FRASER broth with antibiotics
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41106213
NACRES:
NA.74
Recommended Products
Agency
APHA
ISO 11290
Quality Level
sterility
non-sterile
form
powder
manufacturer/tradename
EcoCult®
technique(s)
microbiological culture: suitable
application(s)
food and beverages
microbiology
pathogen testing
storage temp.
10-30°C
suitability
Listeria spp. (spp.)
General description
EcoCult® Half FRASER (Demi FRASER) broth has high nutrient content and a large buffer capacity that creates optimum growth conditions for Listeria spp. Lithium chloride, nalidixic acid, and acriflavine hydrochloride largely inhibit the growth of accompanying bacteria. The detection of the β-D-glucosidase activity is possible by the addition of aesculin and ammonium iron (III) citrate, forming a black complex of aesculin with iron (III) ions. A black coloration can develop during incubation, but this reaction is not exclusive to Listeria spp. and cultures that show no blackening cannot be assumed to be free of Listeria spp. According to EN ISO 11290-1, every primary and secondary enrichment in Fraser broth has to be sub-cultured on selective plating media. The EcoCult® Half FRASER (Demi FRASER) Broth with antibiotics already contains the antibiotics, so it is only necessary to add the Ammonium iron (III) citrate (1.00092) for the preparation of the primary selective enrichment Half Fraser broth. For the secondary enrichment step, Fraser broth can be prepared by the addition of one additional vial reconstituted FRASER Listeria selective supplements (1.00093) per 500 mL prepared Half Fraser Broth.
Application
EcoCult® Half FRASER (Demi FRASER) broth with antibiotics is used for the primary selective enrichment of Listeria spp. (including L. monocytogenes) from food and animal feed as well as from environmental samples in the area of food production and food handling and other materials.
Features and Benefits
- Compliance to major regulatory standards ISO, FDA-BAM, EPA or USP
- Clear labeling and Certificate of Analysis to check compliance at a glance
- Reliable and accurate quality in powder format
- Cost-effective pricing
Legal Information
ECOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service