Skip to Content
Merck

1.37156

SAFC

Pyridoxine hydrochloride

EMPROVE® EXPERT, Ph. Eur., BP, JP, USP, FCC

Pharma Manufacturing

Synonym(s):

Pyridoxine hydrochloride, Adermine hydrochloride, PN HCl, Pyridoxol hydrochloride, Vitamin B6 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · HCl
CAS Number:
Molecular Weight:
205.64
Beilstein:
3632435
MDL number:
UNSPSC Code:
41116107

Agency

BP
JP
Ph. Eur.
USP

Quality Level

reg. compliance

FCC

product line

EMPROVE® EXPERT

mp

214-215 °C (lit.)

application(s)

clinical nutrition
liquid formulation
parenterals
pharmaceutical
vitamins, nutraceuticals, and natural products

storage temp.

15-25°C

SMILES string

Cl[H].Cc1ncc(CO)c(CO)c1O

InChI

1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H

InChI key

ZUFQODAHGAHPFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For protein-based therapeutics, such as monoclonal antibodies (mAbs) or plasma derived proteins, often highly concentrated protein formulations are required to meet intended dosing regimens. However, this usually comes with elevated viscosities and presents a challenge in the manufacturing process and final formulation. Our Viscosity Reduction Excipient Platform reduces the viscosity and helps overcome challenges associated with highly concentrated protein solutions.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation.
Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

The Viscosity Reduction Platform is based on the combination of two new excipients, one amino acid component with one anionic component, that together can better reduce protein viscosity beyond levels achieved with single excipients. Also, combining the two components enables a balance of viscosity reduction and protein stabilization. Pyridoxine hydrochloride Emprove® Expert is part of this new proprietary technology platform. Also, Pyridoxine hydrochloride (Vitamin B6) is used in clinical nutrition for treatment of vitamin B6 deficiency, anemia, mental retardation, Parkinson′s syndrome, gestational diabetes, tetanus and atherosclerosis.

Packaging

1.37156.0100 / 100g / PE bottle
1.37156.1000 / 1kg / PE bottle
1.37156.9012 / 12kg / PE bucket

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service