1.24069
Parteck® TA (Calcium carbonate)
EMPROVE® ESSENTIAL, Ph. Eur., BP, USP, E170, FCC
Pharma Manufacturing
Synonym(s):
Calcium carbonate
About This Item
Recommended Products
grade
GMO free
Halal
Kosher
cGMP
Quality Level
Agency
BP
Ph. Eur.
USP
reg. compliance
FCC
product line
EMPROVE® ESSENTIAL
form
powder
density
2.93 g/mL at 25 °C (lit.)
application(s)
pharmaceutical
solid formulation
foreign activity
Aflatoxin
storage temp.
2-30°C
SMILES string
[Ca++].[O-]C([O-])=O
InChI
1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2
InChI key
VTYYLEPIZMXCLO-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Features and Benefits
- A mineral, nature-identical pigment
- Excellent compatibility with HPMC and PVA film coatings (including our PARTECK® COAT excipient)
- High process efficiency due to lower viscosity of spraying liquids
- Enables an appealing and uniform tablet appearance
- Enhanced opacity due to tailored particle size distribution and unique morphology
Legal Information
Application
also commonly purchased with this product
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service