126625
Bovine Serum Albumin
30% Aqueous Solution, Stabilizer-Free
Synonym(s):
Albumin, Bovine Serum, 30% Aqueous Solution, Stabilizer-Free
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Product Name
Albumin, Bovine Serum, 30% Aqueous Solution, Stabilizer-Free,
description
Merck USA index - 14, 8468
Assay
≥96% (cellulose acetate electrophoresis)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
technique(s)
nuclear extraction: suitable
shipped in
ambient
storage temp.
2-8°C
General description
Albumin, Bovine Serum, 30% Aqueous Solution, Stabilizer-Free is a highly purified preparation, suitable for a variety of in vitro assays.
Bovine serum albumin (BSA) is a non-glycosylated globular, α-helical protein. It consists of 17-disulfide bonds with three domains and two sub-domains each.
Application
Albumin, Bovine Serum, 30% Aqueous Solution, Stabilizer-Free has been used as a component of buffer in whole-cell binding assay and membrane binding assay.
Biochem/physiol Actions
Bovine Serum Albumin (BSA) acts as a blocking agent in enzyme-linked immunosorbent assay (ELISA). It is very crucial in cell culture media and stimulates embryonic stem cells (hESC) differentiation. BSA also helps the transportation of drugs, hormones, and fatty acids.
Packaging
Please refer to vial label for lot-specific concentration.
Warning
Toxicity: Standard Handling (A)
Physical form
30% aqueous solution, sterile-filtered, 0.1% NaN₃, pH 7.2.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Human striatal glia differentially contribute to AD- and PD-specific neurodegeneration.
Xu, et al.
Nature aging, 3, 346-365 (2023)
Kithsiri B Herath et al.
The Journal of antibiotics, 58(11), 686-694 (2006-02-10)
The chemokine receptor, CCR2, is predominantly expressed on monocytes/macrophages, and on a subset of memory T cells. It binds to several CC type chemokines of the monocyte chemoattractant protein (MCP) family of which MCP-1 exhibits the highest affinity. CCR2/MCP-1 expression/association
S Chodankar et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 77(3 Pt 1), 031901-031901 (2008-06-04)
Small-angle neutron scattering (SANS) and dynamic light scattering (DLS) have been used to study conformational changes in protein bovine serum albumin (BSA) due to perturbation in its native structure as induced by varying temperature and pressure, and in presence of
Yuhong Xiao et al.
Journal of immunological methods, 384(1-2), 148-151 (2012-06-27)
The enzyme-linked immunosorbent assay (ELISA) is an extremely common and powerful laboratory technique for detecting proteins by antibodies. Researchers frequently use bovine serum albumin (BSA) as a blocking agent to prevent non-specific binding of antigens and antibodies to the microtiter
Geoffrey L Francis
Cytotechnology, 62(1), 1-16 (2010-04-08)
Albumin has a long historical involvement in design of media for the successful culture of mammalian cells, in both the research and commercial fields. The potential application of albumins, bovine or human serum albumin, for cell culture is a by-product
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service