1.19254
Chlorine Powder Packs for photometers of other manufacturers for 10-ml samples
photometric, 0 - 2.00 mg/l Cl₂ (free) Spectroquant®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116116
NACRES:
SE.11
Recommended Products
Quality Level
usage
sufficient for 100 tests
specific analyte(s)
chlorine
measuring range
0-2.0 mg/L (Cl2)
technique(s)
photometry: suitable
pH
6.3 (20 °C, 10 g/L in H2O)
compatibility
for use with photometers from other manufacturers
detection method
photometric (DPD)
storage temp.
15-25°C
General description
(free chlorine) Method: photometric, DPD 0 - 2.00 mg/l Cl₂ Spectroquant® Chlorine Powder Packs for photometers of other manufacturers for 10-ml samples.
We offer Spectroquant® test kits for photometers of other manufactures. They work seamlessly with other photometer brands. The tests work with the original programming data installed by the manufacturers according to the instrument’s operation manual.
Furthermore, we provide test-specific data for easy programming for each Spectroquant® test kits with any photometer.
We offer Spectroquant® test kits for photometers of other manufactures. They work seamlessly with other photometer brands. The tests work with the original programming data installed by the manufacturers according to the instrument’s operation manual.
Furthermore, we provide test-specific data for easy programming for each Spectroquant® test kits with any photometer.
Legal Information
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service