Skip to Content
Merck
All Photos(6)

Documents

1.17246

Supelco

Reflectometer RQflex® 20

pkg of 1 unit, Reflectoquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NB.35

product line

Reflectoquant®

Quality Level

packaging

pkg of 1 unit

detection method

reflectometric

General description

Read out Reflectoquant® test strips with the portable RQFlex® 20 reflectometer. The successor of the RQFlex® 10 features a redesigned intuitive user interface paired with high accuracy. It allows for fast and easy measurement of a broad range of chemical parameters in the food, water, and environmental applications.

Packaging

Fibre case

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service