1.16977
Ammonium Test Strips, reflectometric
20-180 mg/L (NH₄⁺), REFLECTOQUANT®, for use with RQflex® 20
Synonym(s):
Ammonium chemical test strips, Ammonium test strip for instrument reader
About This Item
Recommended Products
Product Name
Ammonium Test, reflectometric, 20-180 mg/L (NH4+), for use with REFLECTOQUANT®
description
quantitative
Quality Level
specific analyte(s)
ammonium
measuring range
20-180 mg/L (NH4+)
compatibility
RQflex® 20
for use with REFLECTOQUANT®
detection method
reflectometric
storage temp.
15-25°C
General description
Application
- Evaluation of Water Ammonium Ion Test Kit and Its Feasibility for the Analysis of Protein in Food.: This study evaluates the efficacy of an ammonium ion test kit for water quality analysis and its potential application in determining protein content in food samples, showcasing its versatility in both environmental and food sciences (Wu D et al., 2022).
- Technical note: Evaluation of a colorimetric point-of-care test for measuring urine ammonium concentration in periparturient dairy cattle.: This research highlights the use of a colorimetric ammonium test for monitoring the health and metabolic status of dairy cattle, demonstrating its practical application in veterinary medicine (Megahed AA, Constable PD, 2020).
- Using Cellulolytic Nitrogen Fixing Bacterium, Azomonas agilis for Effective Degradation of Agricultural Residues.: Ammonium tests are used to monitor nitrogen fixation and ammonia production in microbial degradation studies, emphasizing their role in sustainable agriculture and waste management (Latt ZK et al., 2018).
- [Examination of drugs and poisons in institutions not equipped with an analyzer of them and poisons--focusing on the simple qualitative test of surfactants].: This study demonstrates the use of simple ammonium tests for the qualitative analysis of surfactants in forensic and toxicological investigations, highlighting their importance in settings lacking advanced analytical instruments (Kojima Y et al., 2003).
- [15N] ammonium test for liver function diagnosis.: The [15N] ammonium test is utilized to assess liver function by monitoring nitrogen metabolism, providing valuable insights into liver health and the impact of various treatments on hepatic function (Jung K et al., 1989).
Analysis Note
Assay
- standard 1:
36 - 44 mg/l
- standard 2: 90 - 110 mg/l
- standard 3: 135 - 165 mg/l
Legal Information
Disclaimer

Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service