Skip to Content
Merck
All Photos(1)

Documents

1.14540

Supelco

COD Test, photometric

photometric, 10-150 mg/L (COD), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

COD Cell Test, photometric, 10-150 mg/L (COD), Spectroquant®

product line

Spectroquant®

Quality Level

analyte functional class(es)

chemical oxygen demand (COD)

measuring range

10-150 mg/L (COD)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric

storage temp.

15-25°C

General description

Method: Photometric, 10 - 150 mg/L, Spectroquant®
The method corresponds to DIN ISO 15705 and is analogous to EPA 410.4, APHA 5220 D, and ASTM D1252-06 B.
The Chemical Oxygen Demand (COD) is a measure for the amount of oxygen consumed during the oxidation of organic matter, which gives a clear indication for the quality of a water source, it is for sure one of the most important parameters in (waste) water analysis. Our Spectroquant® COD Tests allow you to accurately quantify the of COD levels in your sample.
Select the right measurement range for your application needs from our wide range of Spectroquant® COD Tests.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode), which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service