Skip to Content
Merck
All Photos(1)

Key Documents

1.00688

Supelco

BOD Nutrient-Salt Mixture

12 sufficient for 1 L, Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

Product Name

BOD Nutrient-Salt Mixture, 12 sufficient for 1 L, Spectroquant®

product line

Spectroquant®

Quality Level

form

solid

usage

12 sufficient for 1 L

analyte functional class(es)

biochemical oxygen demand (BOD)

technique(s)

photometry: suitable

storage temp.

15-25°C

General description

(with allyl thiourea) for 12 x 1l nutrient-salt solution Spectroquant®
The BOD (biochemical oxygen demand) is defined as that mass of oxygen that is consumed by microorganisms to oxidatively degrade the organic substances in a sample. Using a cell test allows for an easy and efficient determination of BOD, following a modified Winkler method. All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

The Spectroquant® Cell Test for BOD comes with three empty 16 mm round cells as well as all the required reagents to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Application


  • Integrated leachate management approach incorporating nutrient recovery and removal.: This research highlights the use of BOD nutrient-salt mixtures in an integrated leachate management system. The study demonstrates how this approach can effectively recover and remove nutrients from leachate, enhancing the sustainability and efficiency of waste management practices. The findings are significant for environmental chemistry and waste treatment processes (Podder et al., 2020).

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service