Skip to Content
Merck
All Photos(1)

Documents

1.10783

Supelco

Spectromelt® A 10

(di-lithium tetraborate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B4Li2O7
CAS Number:
Molecular Weight:
169.12
MDL number:
UNSPSC Code:
12352302
EC Index Number:
234-514-3
NACRES:
NA.21

grade

for X-ray fluorescence analysis

Quality Level

Assay

≥99.0% (acidimetric)

form

solid

pH

9.1 (20 °C, 100 g/L in H2O, slurry)

mp

930 °C

solubility

141.2 g/L

density

2.35 g/cm3 at 20 °C

bulk density

330 kg/m3

anion traces

chloride (Cl-): ≤50 ppm
phosphate (PO43-): ≤20 ppm
silicate (as SiO2): ≤100 ppm
sulfate (SO42-): ≤50 ppm

cation traces

Ag: ≤5 ppm
Al: ≤10
Ba: ≤5 ppm
Ca: ≤20 ppm
Cd: ≤5 ppm
Co: ≤5 ppm
Cr: ≤5 ppm
Cu: ≤5 ppm
Fe: ≤10 ppm
Ga: ≤5 ppm
K: ≤10 ppm
Mg: ≤5 ppm
Mn: ≤5 ppm
Na: ≤20 ppm
Ni: ≤5 ppm
Pb: ≤5 ppm
Sr: ≤5 ppm
Zn: ≤5 ppm

storage temp.

2-30°C

InChI

1S/4B.2Li.7H2O/h;;;;;;7*1H2

InChI key

DPYKRXYVXDYLEY-UHFFFAOYSA-N

Linkage

Replaces: 10783; 10783-3

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Chloride (Cl): ≤ 50 ppm
Phosphate (PO₄): ≤ 20 ppm
Silicate (SiO₂): ≤ 100 ppm
Sulfate (SO₄): ≤ 50 ppm
Ag (Silver): ≤ 5 ppm
Al (Aluminium): ≤ 10 ppm
Ba (Barium): ≤ 5 ppm
Ca (Calcium): ≤ 20 ppm
Cd (Cadmium): ≤ 5 ppm
Co (Cobalt): ≤ 5 ppm
Cr (Chromium): ≤ 5 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 10 ppm
Ga (Gallium): ≤ 5 ppm
K (Potassium): ≤ 10 ppm
Mg (Magnesium): ≤ 5 ppm
Mn (Manganese): ≤ 5 ppm
Na (Sodium): ≤ 20 ppm
Ni (Nickel): ≤ 5 ppm
Pb (Lead): ≤ 5 ppm
Sr (Strontium): ≤ 5 ppm
Zn (Zinc): ≤ 5 ppm
Loss on ignition (1000 °C): ≤ 1.0 %
Bulk density: about 50

Legal Information

SPECTROMELT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service