Skip to Content
Merck
All Photos(2)

Key Documents

254304

Sigma-Aldrich

Lithium tetraborate

≥99.995% trace metals basis

Synonym(s):

Dilithium tetraborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2B4O7
CAS Number:
Molecular Weight:
169.12
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder
solubility:
H2O: slightly soluble(lit.)

Quality Level

Assay

≥99.995% trace metals basis

form

powder

impurities

≤50.0 ppm Trace Metal Analysis

mp

760-880 °C (lit.)

solubility

H2O: slightly soluble(lit.)

density

≥0.25 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Li+].[Li+].[O-]B(OB=O)OB([O-])OB=O

InChI

1S/B4O7.2Li/c5-1-9-3(7)11-4(8)10-2-6;;/q-2;2*+1

InChI key

PSHMSSXLYVAENJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium tetraborate in combination with oxalic acids and lithium carbonate in the ratio (1:1:1) may be used as an effective fusing agent for siliceous samples. It is widely employed in lithium tetraborate fusion procedure, suited for a variety of soils and rocks. Lithium tetraborate also finds applications in radiation detectors and dosimeters.

Application

Lithium tetraborate may be used as an analytical reagent for the analysis of major and trace elements in coal fly ashes and environmental materials using inductively coupled plasma atomic emission spectrometry (ICP-AES). It may also be used as an alkali fusion reagent for the determination of silicon in zirconium oxide powder using ICP-AES.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of siliceous materials and coal by atomic absorption spectrophotometry with fusion for sample decomposition
Lau W-O, et al.
Talanta, 51(5), 1009-1018 (2000)
Contamination-free decomposition of zirconium oxide for the determination of ultra-trace silicon by inductively coupled plasma-atomic emission spectrometry
Watanabe M, et al.
Analytica Chimica Acta, 416(1), 117-119 (2000)
Analysis of coal fly ash and environmental materials by inductively coupled plasma atomic emission spectrometry: comparison of different decomposition procedures
Bettinelli M, et al.
Journal of Analytical Atomic Spectrometry, 2(5), 485-489 (1987)
Development of a qualitative, multiplex real-time PCR kit for screening of genetically modified organisms (GMOs)
Dorries H-H, et al.
Analytical and Bioanalytical Chemistry, 396(6), 2043-2054 (2010)
Sparks LD, et al.
Methods of Soil Analysis, Part 3: Chemical Methods, 14 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service