Skip to Content
Merck
All Photos(1)

Key Documents

1.06537

Supelco

Sodium nitrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Sodium nitrate, Nitric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NNaO3
CAS Number:
Molecular Weight:
84.99
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-554-3
NACRES:
NA.21
grade:
ACS reagent
form:
solid

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.5% (alkalimetric)

form

solid

potency

1267 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

pH

5.5-8.0 (20 °C, 50 g/L in H2O)

mp

308 °C

density

2.26 g/cm3 at 20 °C

bulk density

1200 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
iodate (IO3-): ≤0.0005%
nitrite (NO2-): ≤0.001%
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.003%

cation traces

Ca: ≤0.002%
Fe: ≤0.0003%
K: ≤0.01%
Mg: ≤0.002%
NH4+: ≤0.002%
heavy metals: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Na+].[N+](=O)([O-])[O-]

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Replaces: SX0655; SX0655-1; SX0655-7

Analysis Note

Assay (alkalimetric): ≥ 99.5 %
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water; 25 °C): 5.5 - 8.0
Chloride (Cl): ≤ 0.0005 %
Iodate: ≤ 0.0005 %
Nitrite (NO₂): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.003 %
Heavy metals (ACS): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.002 %
Fe (Iron): ≤ 0.0003 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.002 %
NH₄ (Ammonium): ≤ 0.002 %
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service