Skip to Content
Merck
All Photos(1)

Key Documents

1.05403

Millipore

GranuCult® prime EE Broth-MOSSEL (acc. harm. EP/USP/JP)

according to EP, according to USP, according to JP, suitable for microbiology

Synonym(s):

Enterobacteria Enrichment (EE) Broth Mossel, Enterobacteriaceae Enrichment Broth – Mossel, Buffered Glucose-Brilliant Green Bile Broth, E.E. Broth, Enterobacteriaceae Enrichment Broth acc. to Mossel, Mossel Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

according to EP
according to JP
according to USP

Quality Level

sterility

non-sterile

form

granular

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® prime

storage condition

dry media (Keep tightly closed)

technique(s)

microbiological culture: suitable

color

green

pH range

7.0-7.4 (25 °C)

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

Escherichia coli
Pseudomonas spp.
Staphylococcus spp.

General description

GranuCult® prime Enterobacteriaceae Enrichment Broth - Mossel is designed for selective enrichment of bile-tolerant, Gram-negative bacteria from non-sterile pharmaceutical products. D (+) Glucose enhances the growth of all Lactose-positive and -negative Enterobacteriaceae. The undesired, accompanying bacterial flora is completely inhibited by brilliant green and ox bile. The strong buffering capacity of the culture medium prevents the formed acid from killing the culture.

Application

GranuCult® prime Enterobacteriaceae Enrichment Broth - Mossel is suitable for sterility testing of substances, preparations, and products as well as a pre-enrichment broth for the test for specified microorganisms or total viable aerobic count determinations using the MPN method within non-sterile pharmaceutical products.

Features and Benefits

  • GranuCult® offers superior granulated culture media
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Analysis Note

Appearance (clearness): clear
Appearance (colour): green
pH-value (25 °C): 7.0 - 7.4
Typical composition (g/litre): Peptone from Gelatine 10.0; D(+)-Glucose 5.0; Ox bile dried 20.0; Brilliant green 0.015; di-Sodium hydrogen phosphate dihydrate 8.0; Potassium dihydrogen phosphate 2.0.
Growth promotion test in accordance with the harmonised method of EP, USP and JP.
Inoculum on reference medium (Escherichia coli ATCC 8739 (WDCM 00012)): 10 - 100
Inoculum on reference medium (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): 10 - 100
Inoculum on reference medium (Staphylococcus aureus ATCC 6538 (WDCM 00032)): ≥ 100
Growth (Escherichia coli ATCC 8739 (WDCM 00012)): +
Growth (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): +
Growth (Staphylococcus aureus ATCC 6538 (WDCM 00032)): no growth
Incubation: 24 hrs.; 30-35 °C; S. aureus 48 hrs.
The information above is current at this time of publication and is subject to change without notice (except for customers holding a change control agreement with our company). The information/format can be altered at any time and inany way if internal company matters or Standard Regulationsdo say so.

Other Notes

  • The prepared broth is clear and green
  • The pH value at 25°C is in the range of 7.0-7.4.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service