1.01095
Aluminum oxide
(anhydrous (γ-aluminia)), Technipur®
Synonym(s):
Alumina
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
MDL number:
UNSPSC Code:
12352311
NACRES:
NA.21
Recommended Products
Quality Level
pH
5 (100 °C, 50 g/L in H2O, suspension)
bp
>2980 °C/1013 hPa
mp
2040 °C (lit.)
SMILES string
O=[Al]O[Al]=O
InChI
1S/2Al.3O
InChI key
TWNQGVIAIRXVLR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Aluminum oxide is an amphoteric oxide of aluminum. It is also known as alumina. It is widely used in abrasives, refractories, electronics, heat-resistant fibers, ceramics, and glass industries due to its wide availability, high thermal stability, and melting point as well as excellent strength and hardness. Alumina distinguishes itself from many composite materials due to its outstanding mechanical, chemical, and thermal properties.
Application
Aluminum oxide (Al2O3) is widely used as an electrical insulator because of its high ionic inter-atomic bonds, excellent thermal resistive properties, and chemical stability. In electronic and microelectronic applications, Al2O3 is used as a substrate for integrated circuits, as well as high-temperature and corrosive-resistant coating materials. Being a dielectric material,Al2O3 possesses a high breakdown strength (300–700 MV m-1) and relatively large bandgap (∼ 9Ev). Therefore it is a potential candidate for use in dielectric capacitors. It is also used in spark plug insulators, micro-electric substrates, and insulating heatsinks.
In plastic and rubber industries, modified Al2O3 is used as a reinforcing filler to improve the rheological and mechanical properties of the composites.
In plastic and rubber industries, modified Al2O3 is used as a reinforcing filler to improve the rheological and mechanical properties of the composites.
Legal Information
Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ruys AJ
Alumina Ceramics: Biomedical and Clinical Applications (2019)
Element and Processing
Soo-Jin Park S-J & Seo M-K
Interface Science and Technology , 18, 431-499 (2011)
Enhancement of properties of red iron oxide ? aluminium oxide solid solutions anticorrosive pigments
Ahmed NM and Selim MM
Pigment & Resin Technology, 34(5), 256-264 (2005)
Preparation and characterisation of high performance rubber vulcanizates loaded with modified aluminium oxide
El-Sabbagh SH, et al.
Pigment & Resin Technology, 35(3), 119-131 (2006)
A novel and simple aluminium/sol?gel-derived amorphous aluminium oxide multilayer film with high energy density
Su Z, et al.
Journal of Material Chemistry C, 6, 5616-5623 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service