Skip to Content
Merck
All Photos(2)

Documents

642991

Sigma-Aldrich

Aluminum oxide

nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
NA.23

form

dispersion
liquid (suspension)
nanoparticles

concentration

20 wt. % in H2O

particle size

30-60 nm (TEM)

density

1.06 g/mL at 25 °C

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aluminium oxide (Al2O3) is a metal oxide based engineered nanosized material that is used in a variety of industrial applications. It can be prepared by the hydrolysis of the alkoxide of Al2O3 and calcinating the particles in the presence of stabilizing agents.

Application

Al2O3 based nanoparticle can be used for a wide range applications such as ceramics, aerospace, wastewater treatment, abrasives, paints, catalysts, and flooring materials.

Other Notes

No dispersant

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanotechnology in high performance paint coatings
Khanna AS
Asian Journal of Experimental Sciences, 21(2), 25-32 (2008)
Burning of nano-aluminized composite rocket propellants
De Luca LT, et al.
Combustion, Explosion and Shock Waves, 41(6), 680-692 (2005)
Alumina and zirconia acrylate nanocomposites coatings for wood flooring: Photocalorimetric characterization
Landry V, et al.
Progress in Organic Coatings, 61(1), 76-82 (2008)
Size-controlled synthesis of alumina nanoparticles from aluminum alkoxides
Park YK, et al.
Materials Research Bulletin, 40(9), 1506-1512 (2005)
Colloidal behavior of aluminum oxide nanoparticles as affected by pH and natural organic matter
Ghosh S, et al.
Langmuir, 24(21), 12385-12391 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service