Skip to Content
Merck
All Photos(1)

Key Documents

261513

Sigma-Aldrich

N,N-Diethyl-p-phenylenediamine

97%

Synonym(s):

4-Amino-N,N-diethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NC6H4NH2
CAS Number:
Molecular Weight:
164.25
Beilstein:
879361
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.571 (lit.)

bp

115-116 °C/5 mmHg (lit.)

mp

19-21 °C (lit.)

solubility

water: insoluble

density

0.988 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCN(CC)c1ccc(N)cc1

InChI

1S/C10H16N2/c1-3-12(4-2)10-7-5-9(11)6-8-10/h5-8H,3-4,11H2,1-2H3

InChI key

QNGVNLMMEQUVQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The reaction of chlorine and N,N-diethyl-p-phenylenediamine for sensitive square-wave voltammetric detection of chlorine has been studied.

Application

N,N-Diethyl-p-phenylenediamine has been employed as a reference probe to investigate rapid chlorination rate constants by a stopped-flow spectrophotometric competition kinetics method.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

282.2 °F - closed cup

Flash Point(C)

139 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduard Rott et al.
International journal of environmental research and public health, 15(5) (2018-05-08)
Effluent of a municipal wastewater treatment plant (WWTP) was treated on-site with the UV/chlorine (UV/HOCl) advanced oxidation process (AOP) using a pilot plant equipped with a medium pressure UV lamp with an adjustable performance of up to 1 kW. Results
Ahmed Refaat Bakr et al.
Chemosphere, 185, 879-887 (2017-07-28)
Bisphenol A is a chemical with hazardous health effects that is largely used in the manufacture of extensively used products including adhesives, plastics, powder paints, thermal paper and paper coatings, and epoxy resin, and is reported to exist in nature
Marta Llorca et al.
Environmental pollution (Barking, Essex : 1987), 246, 346-356 (2018-12-24)
Antidepressant drugs such as Venlafaxine (VFX) and O-desmethylvenlafaxine (ODMVFX) are emerging contaminants that are commonly detected in aquatic environments, since conventional wastewater treatment plants are unable to completely remove them. They can be precursors of hazardous by-products, such as the
Suntae Lee et al.
Pathogens (Basel, Switzerland), 8(4) (2019-11-07)
F-specific RNA bacteriophages (FRNAPHs) can be used to indicate water contamination and the fate of viruses in wastewater treatment plants (WWTPs). However, the occurrence of FRNAPH strains in WWTPs is relatively unknown, whereas FRNAPH genotypes (GI-GIV) are well documented. This
Norhan Nady et al.
Membranes, 10(9) (2020-09-16)
In this work, the efficiency of a conventional chlorination pretreatment is compared with a novel modified low-fouling polyethersulfone (PES) ultrafiltration (UF) membrane, in terms of bacteria attachment and membrane biofouling reduction. This study highlights the use of membrane modification as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service