1.00088
Chlorine Test, photometric
photometric, 0.010 - 6.00 mg/l Cl₂ for free chlorine: Cl₂-1 and Cl₂-2, for total chlorine: Cl₂-1, Cl₂-2, and Cl₂-3 Spectroquant®
About This Item
Recommended Products
Product Name
Chlorine reagent Cl₂-3 (liquid), photometric, 0.010 - 6.00 mg/l Cl₂ for free chlorine: Cl₂-1 and Cl₂-2, for total chlorine: Cl₂-1, Cl₂-2, and Cl₂-3 Spectroquant®
Quality Level
form
liquid
usage
sufficient for 600 tests
specific analyte(s)
chlorine
measuring range
0.010-6.00 mg/L (Cl2)
technique(s)
photometry: suitable
pH
6.5-7.5 (20 °C in H2O)
bp
100 °C
density
1.01 g/cm3 at 20 °C
compatibility
for use with Spectroquant® Move 100
for use with Spectroquant® Move DC
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600
detection method
photometric (DPD)
storage temp.
15-25°C
General description
Application
- Experimental study on mine water purification mechanism for broken coal and rock masses in the underground reservoir of ecologically vulnerable mining area.: Focuses on the purification processes of mine water in fragile ecological zones, using methods including chlorination, to manage pollution and improve water quality (Wang et al., 2024).
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 2 Oral
Target Organs
Thyroid
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service