Skip to Content
Merck
All Photos(2)

Key Documents

07-724

Sigma-Aldrich

Anti-α-Insulin Receptor Antibody, β subunit

Upstate®, from rabbit

Synonym(s):

CD220 antigen, insulin receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

species reactivity (predicted by homology)

chimpanzee

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... INSR(3643)

General description

The insulin receptor is a tyrosine kinase receptor that when bound to insulin, initiates multiple signal transduction pathways, including JNK, PI 3-kinase, Akt and PKC. Pharmacological intervention of these Insulin R-dependent pathways is of great interest for the treatment of insulin resistance, obesity and diabetes. The Insulin Receptor (IR) is synthesized as a single polypeptide, which is subsequently cleaved to generate an extracellular α-chain and a transmembrane and intracellular β-chain, tethered together by disulfide bonds. The β-chain has multiple tyrosine phosphorylation sites, including three autophosphorylation sites at its activation loop. The overall structure of the IR is highly homologous to the IGF-I Receptor, except in their c-termini, where the two proteins diverge somewhat. The IR signals primarily by phosphorylating the Insulin Receptor Substrate (IRS) family of proteins, which creates docking sites for SH2-domain containing proteins. Insulin signaling is highly dependent on the PI3 Kinase pathway and Akt, which appear to mediate the functions of insulin.

Specificity

This antibody recognizes α-Insulin Receptor, β subunit.

Immunogen

Epitope: C-terminus
GST-fusion protein corresponding to the last 100 C-terminal amino acid residues of the human insulin receptor β chain

Application

Anti-α-Insulin Receptor Antibody, β subunit is an antibody against a-Insulin Receptor for use in IP & WB.
Immunoprecipitation Analysis:
Research Category
Signaling

Metabolism
Research Sub Category
Insulin/Energy Signaling

Quality

Evaluated by Western Blot in Human brain tissue lysate.
Western Blot Analysis: : 1:1,000 dilution of this antibody detected α-Insulin Receptor, β subunit in human brain tissue lysate.

Target description

97 kDa

Physical form

Affinity purified
Purified rabbit polyclonal serum in buffer containing 0.1 M Tris-glycine, pH 7.4, 150 mM NaCl and 0.05% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C for for 1 year from date of receipt.

Analysis Note

Control
Human brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin receptor and IRS-1 co-immunoprecipitation with SOCS-3, and IKK?/? phosphorylation are increased in obese Zucker rat skeletal muscle.
Zolotnik, IA; Figueroa, TY; Yaspelkis, BB
Life Sciences null
Makoto Takahashi et al.
Oncology reports, 37(5), 2929-2941 (2017-04-11)
We investigated the relationship among serum insulin level, insulin receptor (IR) expression in renal cell carcinoma (RCC), and outcomes in patients with RCC who underwent nephrectomy. We also explored the role of insulin signaling in RCC progression in a murine RCC
Insulin responsiveness in metabolic syndrome after eight weeks of cycle training.
Stuart, CA; South, MA; Lee, ML; McCurry, MP; Howell, ME; Ramsey, MW; Stone, MH
Med Sci Sports Exerc. null
Michael E Cox et al.
The Prostate, 69(1), 33-40 (2008-09-12)
Although recent laboratory and population studies suggest that prostate cancer may be responsive to insulin, there is a gap in knowledge concerning the expression of insulin receptors on benign or malignant prostate tissue. We immunostained 644 cores on tissue microarrays
Redistribution of glycolipid raft domain components induces insulin-mimetic signaling in rat adipocytes.
Muller, G, et al.
Molecular and cellular biology, 21, 4553-4567 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service