Skip to Content
Merck
All Photos(8)

Documents

07-030

Sigma-Aldrich

Anti-dimethyl-Histone H3 (Lys4) Antibody

Upstate®, from rabbit

Synonym(s):

H3K4me2, Histone H3 (di methyl K4), H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

Tetrahymena sp., human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

dimethylation (Lys4)

Gene Information

human ... H3C1(8350)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Broad species cross-reactivity is expected.
Recognizes dimethylated Histone H3 (Lys4), MW ~17 kDa. Specificity demonstrated by ELISA using methylated peptides.

Immunogen

Epitope: dimethylated Histone H3 (Lys4)
KLH conjugated synthetic peptide (ARTMe2KQTAR-GC) corresponding to amino acids 1-8 of human Histone H3.

Application

Chromatin Immunoprecipitation:
5 μL of a previous lot immunoprecipitated methylated Histone H3 from 1 x 106 3T3/A31 cells.

Immunocytochemistry:
1:500 dilution of a previous lot showed positive immunostaining for methylated Histone H3 in L6 cells fixed with 4% paraformaldehyde and permeabilized using 0.1% Triton X-100.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-dimethyl-Histone H3 (Lys4) Antibody (rabbit polyclonal antibody) is published and validated in ChIP, DB, ICC, IF, WB, PIA, ChIP-seq. to detect dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4).

Quality

Evaluated by western blot on HeLa acid extract.

Western Blot Analysis: 1:2000-1:10,000 dilution of this antibody detected methylated Histone H3 in acid extracted proteins from HeLa cells.
1:2000-1:5000 dilution of a previous lot detected methylated Histone H3 in acid extracted proteins from HeLa and tetrahymena cells.

Target description

Approx. 17 kDa

Physical form

Format: Unpurified
Rabbit polyclonal antiserum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa acid extract

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The requirements for COMPASS and Paf1 in transcriptional silencing and methylation of histone H3 in Saccharomyces cerevisiae.
Mueller, JE; Canze, M; Bryk, M
Genetics null
Dorota Skowronska-Krawczyk et al.
Development (Cambridge, England), 131(18), 4447-4454 (2004-09-03)
Basic helix-loop-helix (bHLH) transcription factors such as atonal homolog 5 (ATH5) and neurogenin 2 (NGN2) determine crucial events in retinogenesis. Using chromatin immunoprecipitation, we demonstrate that their interactions with target promoters undergo dynamic changes as development proceeds in the chick
Epigenetic changes to the MDR1 locus in response to chemotherapeutic drugs.
Baker, EK; Johnstone, RW; Zalcberg, JR; El-Osta, A
Oncogene null
Pharmacologic inhibition of epigenetic modifications, coupled with gene expression profiling, reveals novel targets of aberrant DNA methylation and histone deacetylation in lung cancer.
Zhong, S; Fields, CR; Su, N; Pan, YX; Robertson, KD
Oncogene null
Distribution of histone H3.3 in hematopoietic cell lineages.
Jin, C; Felsenfeld, G
Proceedings of the National Academy of Sciences of the USA null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service