Skip to Content
Merck
All Photos(1)

Key Documents

07-631

Sigma-Aldrich

Anti-cdk2 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-CDKN2, Anti-p33(CDK2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CDK2AP1(8099)

Specificity

Predicted to cross-react with mouse, rat, non-human primates, and hamster based on sequence homology
Recognizes human p33cdk2 protein kinase.

Immunogen

peptide corresponding to amino acids 287-298 (C-QDVTKPVPHLRL) of human cdk2

Application

Anti-cdk2 Antibody is a high quality Rabbit Polyclonal Antibody for the detection of cdk2 & has been validated in IP & WB.

Quality

routinely evaluated by immunoblot in nuclear extracts from HeLa cells

Target description

33 kDa

Physical form

Format: Purified

Analysis Note

Control
Colon tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sublytic complement attack induces cell cycle in oligodendrocytes.
Rus, H G, et al.
Journal of immunology (Baltimore, Md. : 1950), 156, 4892-4900 (1996)
S J Elledge et al.
The EMBO journal, 10(9), 2653-2659 (1991-09-01)
The onset of S-phase and M-phase in both Schizosaccharomyces pombe and Saccharomyces cerevisiae requires the function of the cdc2/CDC28 gene product, p34, a serine-threonine protein kinase. A human homolog, p34cdc2, was identified by functional complementation of the S.pombe cdc2 mutation
Carlotta Costa et al.
Cancer discovery, 10(1), 72-85 (2019-10-09)
The combination of CDK4/6 inhibitors with antiestrogen therapies significantly improves clinical outcomes in ER-positive advanced breast cancer. To identify mechanisms of acquired resistance, we analyzed serial biopsies and rapid autopsies from patients treated with the combination of the CDK4/6 inhibitor
CDK2 encodes a 33-kDa cyclin A-associated protein kinase and is expressed before CDC2 in the cell cycle.
Elledge, S J, et al.
Proceedings of the National Academy of Sciences of the USA, 89, 2907-2911 (1992)
Dissecting the role of p53 phosphorylation in homologous recombination provides new clues for gain-of-function mutants.
Restle, A; Farber, M; Baumann, C; Bohringer, M; Scheidtmann, KH; Muller-Tidow, C; Wiesmuller, L
Nucleic Acids Research null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service