Skip to Content
Merck
All Photos(3)

Key Documents

06-570-AF488

Sigma-Aldrich

Anti-phospho Histone H3 (Ser10) Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

H3S10P, Histone H3 (phospho Ser10), H3 histone, family 3A, H3.3A, H3 histone, family 3B, H3.3B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer10)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Immunogen

Linear peptide corresponding to phospho-Histone H3 (Ser10).

Application

Anti-phospho-H3 (Ser10), Alexa Fluor 488 Conjugate is a purified rabbit polyclonal that detects Histone H3 phosphorylated at serine 10. This Ab is labeled with Alexa Fluor 488 and validated in IC. The unconjugated version of this antibody is highly published and proven in ICC, IP & WB.
Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected phospho Histone H3 (Ser10) in NIH/3T3 cells.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Histone H3 (Ser10) in HeLa cell lysate.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:50 dilution of this antibody detected phospho Histone H3 (Ser10) in HeLa cells.

Alexa Fluor is a registered trademark of Life Technologies.

Target description

~17 kDa observed

Physical form

Protein A purified
Purified rabbit conjugate in buffer containing PBS, 15 mg/ml BSA, and 0.1% Sodium Azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baiping Cui et al.
Cell regeneration (London, England), 11(1), 9-9 (2022-04-02)
Myocardial regeneration has been considered a promising option for the treatment of adult myocardial injuries. Previously, a chick early amniotic fluid (ceAF) preparation was shown to contain growth-related factors that promoted embryonic growth and cellular proliferation, though the nature of
Edward E Large et al.
PLoS genetics, 13(5), e1006769-e1006769 (2017-05-12)
Most biological traits and common diseases have a strong but complex genetic basis, controlled by large numbers of genetic variants with small contributions to a trait or disease risk. The effect-size of most genetic variants is not absolute and is
Xinlu Gao et al.
International journal of medical sciences, 19(8), 1254-1264 (2022-08-06)
Mammalian cardiomyocytes (CMs) maintain a low capacity for self-renewal in adulthood, therefore the induction of CMs cycle re-entry is an important approach to promote myocardial repair after injury. Recently, photobiomodulation (PBM) has been used to manipulate physiological activities of various
Gaetana Gambino et al.
Open biology, 12(12), 220216-220216 (2022-12-22)
Cell quiescence appeared early in evolution as an adaptive response to adverse conditions (i.e. nutrient depletion). In metazoans, quiescence has been involved in additional processes like tissue homeostasis, which is made possible by the presence of adult stem cells (ASCs).
Erin J Vanzyl et al.
PloS one, 13(1), e0191178-e0191178 (2018-01-18)
The spliceosome is a large ribonucleoprotein complex that catalyzes the removal of introns from RNA polymerase II-transcribed RNAs. Spliceosome assembly occurs in a stepwise manner through specific intermediates referred to as pre-spliceosome complexes E, A, B, B* and C. It

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service