Skip to Content
Merck

05-806-AF647

Anti-phospho Histone H3 (Ser10), clone 3H10, Alexa Fluor™ 647 Antibody

clone 3H10, from mouse, ALEXA FLUOR™ 647

Synonym(s):

H3S10P, Histone H3 (phospho Ser10), H3 histone, family 3A, H3.3A, H3 histone, family 3B, H3.3B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
ALEXA FLUOR™ 647
Clone:
3H10, monoclonal
Application:
ICC
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

ALEXA FLUOR™ 647

antibody form

purified antibody

antibody product type

primary antibodies

clone

3H10, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer10)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Histone proteins are highly post-translationally modified, and Histone H3 is the most extensively modified of the five histones. Histone H3 sequence variants and variable modification states are thought to play a role in the dynamic and long term regulation of genes. Di- and tri-methylation of histone H3 are gene silencing marks, while removal of these marks by the demethylase genes Jmjd1a and Jmjd2c leads to gene activation. This product is a conjugated version of Cat. No. 05-806.
~17 kDa observed (Refer to Cat. No. 05-806)

Immunogen

Linear peptide corresponding to Histone H3 phosphorylated at Ser10.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-phospho Histone H3 (Ser10) antibody, clone 3H10, Alexa Fluor™ 647 is a purified mouse monoclonal for detection of Histone H3 phosphorylated at serine 10.
This mAb is fluorescently labeled and validated for use in IC. The unconjugated version has been proven in FC, ICC, IF, PIA, WB, Mplex.

Physical form

Protein G Purified
Purified mouse conjugated antibody in buffer containing PBS and 15 mg/ml BSA with 0.1% Sodium Azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Immunocytochemistry Analysis in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected phospho Histone H3 (Ser10) in HeLa cells.

Alexa Fluor™ is a registered trademark of Life Technologies.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library