Skip to Content
Merck
All Photos(1)

Key Documents

05-725

Sigma-Aldrich

Anti-53BP1 Antibody, clone BP18

ascites fluid, clone BP18, Upstate®

Synonym(s):

Anti-Anti-53BP1, Anti-Anti-TDRD30, Anti-Anti-p202, Anti-Anti-p53BP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

BP18, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TP53BP1(7158)
mouse ... Trp53Bp1(27223)

Specificity

53BP1

Immunogen

Mix of three GST fusion proteins corresponding to residues 1-337, 338-671, and 1331-1664, respectively, of human 53BP1

Application

Detect 53BP1 with Anti-53BP1 Antibody, clone BP18 (Mouse Monoclonal Antibody), that has been shown to work in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

routinely evaluated by immunoblot on whole cell lysates from HeLa cells

Target description

~250kDa

Physical form

Ascites
mouse ascites IgM containing 0.05% sodium azide and 30% glycerol

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Pinkerneil et al.
Targeted oncology, 11(6), 783-798 (2016-06-03)
Targeting of class I histone deacetylases (HDACs) exerts antineoplastic actions in various cancer types by modulation of transcription, upregulation of tumor suppressors, induction of cell cycle arrest, replication stress and promotion of apoptosis. Class I HDACs are often deregulated in
Irene M Ward et al.
Molecular and cellular biology, 23(7), 2556-2563 (2003-03-18)
53BP1 is a p53 binding protein of unknown function that binds to the central DNA-binding domain of p53. It relocates to the sites of DNA strand breaks in response to DNA damage and is a putative substrate of the ataxia
Maria Pinkerneil et al.
Molecular cancer therapeutics, 15(2), 299-312 (2016-01-17)
Class I histone deacetylases HDAC1 and HDAC2 contribute to cell proliferation and are commonly upregulated in urothelial carcinoma. To evaluate whether specific inhibition of these enzymes might serve as an appropriate therapy for urothelial carcinoma, siRNA-mediated knockdown and specific pharmacologic
I Rappold et al.
The Journal of cell biology, 153(3), 613-620 (2001-05-02)
The tumor suppressor p53 binding protein 1 (53BP1) binds to the DNA-binding domain of p53 and enhances p53-mediated transcriptional activation. 53BP1 contains two breast cancer susceptibility gene 1 COOH terminus (BRCT) motifs, which are present in several proteins involved in
Margarita Pustovalova et al.
International journal of molecular sciences, 21(9) (2020-05-14)
Radiation therapy is one of the main methods of treating patients with non-small cell lung cancer (NSCLC). However, the resistance of tumor cells to exposure remains the main factor that limits successful therapeutic outcome. To study the molecular/cellular mechanisms of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service