Skip to Content
Merck
All Photos(1)

Key Documents

05-633

Sigma-Aldrich

Anti-BrdU Antibody, clone BU-1

culture supernatant, clone BU-1, Upstate®

Synonym(s):

Anti-BrdU antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

BU-1, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable

isotype

IgG2a

shipped in

wet ice

target post-translational modification

unmodified

General description

The immunocytochemical detection of bromodeoxyuridine (BrdU) incorporated into DNA is a powerful tool to study the cytokinetics of normal and neoplastic cells. In vitro or in vivo labeling of tumor cells with the thymidine analogue BrdU and the subsequent detection of incorporated BrdU with specific anti-BrdU monoclonal antibodies is an accurate and comprehensive method to quantitate the degree of DNA-synthesis. BrdU is incorporated into the newly synthezised DNA of the S-phase cells and can thus provide an estimate for the fraction of cells in S-phase. Also dynamic proliferative information (such as the S-phase transit rate and the potential doubling time) can be obtained, by means of bivariate BrdU/DNA flow cytometric analysis.

Specificity

5-Bromo-2-deoxyuridine

Immunogen

5-iodouridine:ovalbumin conjugate

Application

Detect BrdU also known as Bromodeoxyuridine with Anti-BrdU Antibody, clone BU-1 (Mouse Monoclonal Antibody), that has been demonstrated to work in ICC.

Quality

A representative lot was evaluated by immunocytochemistry on HeLa cells. A 1:2-1:4 dilution from a representative lot showed positive immunostaining for Anti-BrdU in HeLa cells fixed with 95% ethanol, 5% glacial acetic acid. This lot was tested using the DNA Replication Assay Kit (Catalog # 17-316).

Technical Tip: This antibody detects 5-Bromo-2- deoxyuridine incorporated into double-stranded DNA. DNA denaturation is not required.

Target description

none given

Physical form

4 vials, each vial containing mouse culture supernatant lyophilized from 2.5 mL. The supernatant contains mycoplasma, which contribute enzymes that facilitate antibody binding. Lyophilized powder.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Gao et al.
Cell death & disease, 6, e1610-e1610 (2015-01-23)
G-quadruplex (G4) DNA and G4 DNA resolvase are involved in a variety of biological processes. To understand the biological function of G4 DNA structures and their resolvases in spermatogenesis, we investigated the distribution of G4 structures in mouse testis and
S-phase detection with an antibody to bromodeoxyuridine. Role of DNase pretreatment.
Gonchoroff, N J, et al.
Journal of Immunological Methods, 93, 97-101 (1986)
Chunhua Tang et al.
Journal of molecular cell biology, 11(5), 408-420 (2018-09-15)
DICER1 is a key enzyme responsible for the maturation of microRNAs. Recent evidences suggested that DICER1 and microRNAs expressed in epididymis were involved in the control of male fertility. However, the exact mechanism remains to be elucidated. Here, we created
N J Gonchoroff et al.
Cytometry, 6(6), 506-512 (1985-11-01)
We describe a mouse monoclonal antibody (BU-1) reactive with 5-bromo-2-deoxyuridine (BrdUrd). The antibody is different from previously described BrdUrd monoclonal antibodies in that BU-1 does not require pretreatment of cells with strong DNA denaturants in order for the antibody to
Difference between random and imprinted X inactivation in common voles.
Elena V Dementyeva,Alexander I Shevchenko,Olga V Anopriyenko,Nina A Mazurok et al.
Chromosoma null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service