Skip to Content
Merck
All Photos(4)

Documents

05-516

Sigma-Aldrich

Anti-Ras Antibody, clone RAS10

clone RAS10, Upstate®, from mouse

Synonym(s):

GTP- and GDP-binding peptide B, GTPase Hras, Ha-Ras1 proto-oncoprotein, Ras family small GTP binding protein H-Ras, Transforming protein p2, c-has/bas p21 protein, c-ras-Ki-2 activated oncogene, p19 H-RasIDX protein, transformation gene: oncogene HAMSV,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RAS10, monoclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Related Categories

General description

The tumor oncoproteins HRas, KRas, and NRas are rated as Ras protoncogenes. Point mutations within Ras genes are frequently detected in human malignancies and in experimentally induced tumors in animals.

Specificity

Other species unknown.

Immunogen

Purified recombinant Ras. Clone RAS10.

Application

Detect Ras using this Anti-Ras Antibody, clone RAS10 validated for use in ELISA, FC, IC, IH, IP & WB.
Immunoprecipitation:
1-4 μg of this antibody has been reported to immunoprecipitate Ras.

Immunocytochemistry:
2.5 μg/mL of this antibody has been reported to show positive immunostaining for Ras in SW480 and Y1 cells.

Immunohistochemistry:
This antibody has been reported to detect Ras in frozen and paraffin embedded breast carcinoma sections.

Quality

Routinely evaluated by western blot on RIPA lysate from human A431 carcinoma cells, human HFF, Jurkat, Raji, mouse 3T3, CTLL, rat L-6 or PC-12 cell lysates.

Western Blot Analysis:
0.05 - 0.5 μg/mL of this lot detected Ras in RIPA lysate from human A431 carcinoma cells. A previous lot detected Ras in human HFF, Jurkat, Raji, mouse 3T3, CTLL, rat L-6 and PC- 12 cell lysates.

Target description

21 kDa

Linkage

Replaces: CBL751

Physical form

Format: Purified
Purified mouse monoclonal IgG2ak in buffer containing PBS, pH 7.4, with 0.05% sodium azide and 30% glycerol. Liquid at -20°C.

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ras plasma membrane signalling platforms.
Hancock, John F and Parton, Robert G
The Biochemical Journal, 389, 1-11 (2005)
GTP hydrolysis mechanism of Ras-like GTPases.
Li, Guangpu and Zhang, Xuejun C
Journal of Molecular Biology, 340, 921-932 (2004)
The Ras-MAPK signal transduction pathway, cancer and chromatin remodeling.
Dunn, Katherine L, et al.
Biochemistry and Cell Biology = Biochimie Et Biologie Cellulaire, 83, 1-14 (2005)
Oncogenic Ras and its role in tumor cell invasion and metastasis.
Campbell, Paul M and Der, Channing J
Seminars in cancer biology, 14, 105-114 (2004)
Oncogenic Ras in tumour progression and metastasis.
Giehl, Klaudia
Biological Chemistry, 386, 193-205 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service