Skip to Content
Merck
All Photos(1)

Key Documents

05-206

Sigma-Aldrich

Anti-Laminin beta 2 (LAMB2) Antibody

UPSTATE®, rat monoclonal, A5

Synonym(s):

Anti-LAMS, Anti-NPHS5, Anti-PIERS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Laminin B2 Antibody, clone A5, clone A5, Upstate®, from rat

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

A5, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LAMB2(3913)

Specificity

B2 chain laminin; does not cross-react with other basement membrane components or fibronectin

Immunogen

Murine EHS laminin

Application

Anti-Laminin B2 Antibody, clone A5 is an antibody against Laminin B2 for use in IP, IH.
Not recommended for WB.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

routinely evaluated on unfixed, frozen monkey brain sections

Target description

196 kDa

Physical form

0.1M Tris-glycine, pH 7.0
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbora Malecova et al.
eLife, 5 (2016-02-18)
Change in the identity of the components of the transcription pre-initiation complex is proposed to control cell type-specific gene expression. Replacement of the canonical TFIID-TBP complex with TRF3/TBP2 was reported to be required for activation of muscle-gene expression. The lack
Cdc42 is crucial for the establishment of epithelial polarity during early mammalian development.
Xunwei Wu,Shaohua Li,Anna Chrostek-Grashoff,Aleksandra Czuchra,Hannelore Meyer et al.
Developmental Dynamics null
Wei Liu et al.
Frontiers in molecular neuroscience, 10, 239-239 (2017-08-30)
Background: The cochlea produces an electric field potential essential for hair cell transduction and hearing. This biological "battery" is situated in the lateral wall of the cochlea and contains molecular machinery that secretes and recycles K+ ions. Its functioning depends
Optimization of immunohistochemical techniques to detect extracellular matrix proteins in fixed skin specimens.
Folkvord, J M, et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 37, 105-113 (1989)
Andrzej Steplewski et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 34(3), 489-501 (2015-08-05)
Post-traumatic joint contracture is a debilitating consequence of trauma or surgical procedures. It is associated with fibrosis that develops regardless of the nature of initial trauma and results from complex biological processes associated with inflammation and cell activation. These processes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service