Skip to Content
Merck
All Photos(1)

Documents

Z4152

Sigma-Aldrich

Zafirlukast

≥98% (HPLC)

Synonym(s):

N-[3-[[2-Methoxy-4-[[[(2-methylphenyl)sulfonyl]amino]carbonyl]phenyl]methyl]-1-methyl-1H-indol-5-yl]carbamic acid cyclopentyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H33N3O6S
CAS Number:
Molecular Weight:
575.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: ≥28 mg/mL

originator

AstraZeneca

storage temp.

room temp

SMILES string

COc1cc(ccc1Cc2cn(C)c3ccc(NC(=O)OC4CCCC4)cc23)C(=O)NS(=O)(=O)c5ccccc5C

InChI

1S/C31H33N3O6S/c1-20-8-4-7-11-29(20)41(37,38)33-30(35)22-13-12-21(28(17-22)39-3)16-23-19-34(2)27-15-14-24(18-26(23)27)32-31(36)40-25-9-5-6-10-25/h4,7-8,11-15,17-19,25H,5-6,9-10,16H2,1-3H3,(H,32,36)(H,33,35)

InChI key

YEEZWCHGZNKEEK-UHFFFAOYSA-N

Gene Information

human ... CYSLTR1(10800)

Application

Zafirlukast has been used to stimulate pancreatic β cell line (MIN6) and pancreatic islets for insulin secretion assay. It may be used as an adenosine triphosphate-binding cassette transporter (ABCG2) inhibitor in MTT [3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide] cytotoxicity assay in human embryonic kidney cells (HEK293) and in Kirby-Bauer disc diffusion assays, bactericidal activity and minimal inhibitory concentration (mIC) assay against M. smegmatis

Biochem/physiol Actions

Zafirlukast is a potent subtype specific cysteinyl leukotriene type 1 receptor (CysLT1) antagonist; antiasthmatic. Zafirlukast has over 1000-fold selectivity for CysLT1, one of two receptors for cysteinyl leukotrienes LTC4, LTD4, and LTE4, which are important mediators of human bronchial asthma.
Zafirlukast plays a key role in alleviating mucus and airway constriction in asthma based inflammatory response. It regulates pancreatic β cells for the secretion of insulin and this functionality is interlinked to the calcium based phosphorylation of the Ca2+/calmodulin-dependent protein kinase II (CaMKII) and protein kinase B (AKT). Zafirlukast has inhibitory potential on ATP-binding cassette (ABC) transporters and reverses the multidrug resistance function of ATP-binding cassette super-family G member 2 (ABCG2). Zafirlukast inhibits mycobacterial nucleoid-associated protein Lsr2 and halts the growth of Mycobacteria.

Features and Benefits

This compound is featured on the Leukotriene Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J C Adkins et al.
Drugs, 55(1), 121-144 (1998-02-17)
Zafirlukast is a competitive and selective leukotriene receptor antagonist indicated for the prophylaxis and treatment of chronic asthma. The rationale for the development of leukotriene antagonists was based on in vitro and in vivo data demonstrating the extensive role of
P N Richard Dekhuijzen et al.
Clinical pharmacokinetics, 41(2), 105-114 (2002-03-13)
Zafirlukast is a cysteinyl leukotriene type 1 receptor antagonist that causes bronchodilation and has anti-inflammatory properties. Clinical efficacy has been demonstrated when using oral doses of 20 to 40 mg twice daily. The pharmacokinetics of zafirlukast are best described by
F Wunder et al.
British journal of pharmacology, 160(2), 399-409 (2010-04-29)
Cysteinyl leukotrienes (CysLTs) have been implicated in the pathophysiology of inflammatory and cardiovascular disorders. Their actions are mediated by CysLT(1) and CysLT(2) receptors. Here we report the discovery of 3-({[(1S,3S)-3-carboxycyclohexyl]amino}carbonyl)-4-(3-{4-[4-(cyclo-hexyloxy)butoxy]phenyl}propoxy) benzoic acid (HAMI3379), the first potent and selective CysLT(2) receptor
Lucile Pinault et al.
Antimicrobial agents and chemotherapy, 57(5), 2134-2140 (2013-02-27)
The mycobacterial nucleoid-associated protein Lsr2 is a DNA-bridging protein that plays a role in condensation and structural organization of the genome and acts as a global repressor of gene transcription. Here we describe experiments demonstrating that zafirlukast inhibits the complexation
Tiina Karonen et al.
European journal of clinical pharmacology, 67(2), 151-155 (2010-10-12)
Gemfibrozil, a strong inhibitor of cytochrome P450 (CYP) 2C8 in vivo, was recently found to markedly increase the plasma concentrations of montelukast in humans. Like montelukast, zafirlukast is a substrate of CYP2C9 and CYP3A4 and a potent inhibitor of CYP2C8

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service