Skip to Content
Merck
All Photos(1)

Key Documents

S-015

Supelco

Sirolimus solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Rapamycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H79NO13
CAS Number:
Molecular Weight:
914.17
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

CO[C@@H]1C[C@@H](CC[C@H]1O)C[C@H](C)[C@@H]2CC(=O)[C@H](C)C=C(C)[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(C)[C@H](C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N4CCCC[C@H]4C(=O)O2)OC

InChI

1S/C51H79NO13/c1-30-16-12-11-13-17-31(2)42(61-8)28-38-21-19-36(7)51(60,65-38)48(57)49(58)52-23-15-14-18-39(52)50(59)64-43(33(4)26-37-20-22-40(53)44(27-37)62-9)29-41(54)32(3)25-35(6)46(56)47(63-10)45(55)34(5)24-30/h11-13,16-17,25,30,32-34,36-40,42-44,46-47,53,56,60H,14-15,18-24,26-29H2,1-10H3/b13-11+,16-12+,31-17+,35-25+/t30-,32-,33-,34-,36-,37+,38+,39+,40-,42+,43+,44-,46-,47+,51-/m1/s1

InChI key

QFJCIRLUMZQUOT-HPLJOQBZSA-N

Gene Information

human ... FKBP1A(2280)

General description

This Certified Spiking Solution® is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the drug’s narrow therapeutic range. The Snap-N-Spike® format allows laboratories to eliminate weighing operations of hazardous substances such as sirolimus, significantly reducing occupational exposure hazards related to the handling of acutely toxic powders, and provides the ability to spike into their matrix of choice just before use.

Sirolimus, also known as rapamycin, belongs to the group of natural hydrophobic macrolides and is used widely as an immunosuppressive drug. It also shows anti-fungal, anti-candidal, anti-tumor, and anti-proliferative, in addition to immunosuppressive activities. Sirolimus is produced by Streptomyces hygroscopicus bacterium and works as a mammalian Target of Rapamycin (mTOR)inhibitor. It acts by blocking the activation and proliferation of T-lymphocytes, in response to the cytokines and antigens.

Application

The certified reference solution (CRM) in solution can also be used as follows:
  • Multi-residue analysis of sirolimus, tacrolimus, everolimus, and cyclosporine A in whole blood samples by combining Bio-solid phase microextraction with mass spectrometry through a microfluidic open interface (Bio-SPME-MOI-MS/MS)
  • Development and validation of a high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) to determine immunosuppressive drugs― cyclosporine A, tacrolimus, sirolimus, and everolimus in whole blood samples
  • Determination of sirolimus in both pig whole blood and lung tissue samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) and using a single protein precipitation step for sample preparation
  • Analysis of blood samples collected from former transplant patients for the quantification of sirolimus, cyclosporine A, tacrolimus, and everolimus by liquid chromatography and tandem mass spectrometryEstimation of sirolimus, cyclosporine A, and everolimus in the dried matrix on paper discs (DMPD) by liquid chromatography along with tandem mass spectrometry (LC-MS/MS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Albert et al.
The Journal of cell biology, 202(2), 201-210 (2013-07-24)
Chromosomes architecture is viewed as a key component of gene regulation, but principles of chromosomal folding remain elusive. Here we used high-throughput live cell microscopy to characterize the conformation and dynamics of the longest chromosome of Saccharomyces cerevisiae (XII). Chromosome
M A Beigh et al.
Journal of biological regulators and homeostatic agents, 27(2), 399-408 (2013-07-09)
S6K1 regulation associates a central role with dynamics of sequential phosphorylations at the hydrophobic motif (T412) and activation loop (T252) of the enzyme, such that the hydrophobic motif phosphorylation supposedly brought about by mTOR- kinase, primes the enzyme for PDK1
Beyza Macunluoglu et al.
JPMA. The Journal of the Pakistan Medical Association, 62(8), 822-825 (2013-07-19)
To investigate the effects of the potent immunosuppressive agents tacrolimus and rapamycin on the number of circulating mature endothelial cells and circulating endothelial progenitor cells in an experimental model. It was an experimental study performed from December 2007 to January
Young Joo Shin et al.
Investigative ophthalmology & visual science, 54(7), 4452-4458 (2013-05-30)
To investigate the protective effect of rapamycin against alkali burn-induced corneal damage in mice. BALB/c mice were treated with 0.1 N NaOH to the cornea for 30 seconds. Corneal neovascularization and opacity were clinically evaluated at 1, 2, and 4
Tabetha Sundin et al.
Experimental cell research, 319(13), 2103-2112 (2013-06-12)
Translation is mediated partly by regulation of free eukaryotic initiation factor 4E (eIF4E) levels through PI3K-Akt-mTOR signaling. Cancer cells treated with the plant-derived perillyl alcohol (POH) or the mechanistic target of rapamycin (mTOR) inhibitor rapamycin dephosphorylate eIF4E-binding protein (4E-BP1) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service