Skip to Content
Merck
All Photos(1)

Key Documents

P-069

Supelco

Progesterone solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC(=O)C1CCC2C3CCC4=CC(=O)CCC4(C)C3CCC12C

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1

InChI key

RJKFOVLPORLFTN-LEKSSAKUSA-N

Gene Information

human ... PGR(5241)

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K L Bruner-Tran et al.
Minerva ginecologica, 65(2), 199-213 (2013-04-20)
Progesterone action normally mediates the balance between anti-inflammatory and proinflammatory processes throughout the female reproductive tract. However, in women with endometriosis, endometrial progesterone resistance, characterized by alterations in progesterone responsive gene and protein expression, is now considered a central element
J H Check et al.
Clinical and experimental obstetrics & gynecology, 40(1), 37-39 (2013-06-04)
To determine if pregnancy rates following embryo transfer are reduced if the endometrial echo pattern in the late proliferative phase is isoechogenic (IE) vs triple line (TL). Pregnancy and implantation rates were compared according to TL vs IE pattern in
Ratana Sapbamrer et al.
Asia Pacific journal of clinical nutrition, 22(2), 222-228 (2013-05-03)
Isoflavone in soybean and its products have numerous beneficial health effects. A number of clinical studies have demonstrated that dietary soy isoflavone can relieve menopausal symptoms, lower risks of breast cancer, and lower cholesterol and glucose. Among the various effects
Cathrin Brisken
Nature reviews. Cancer, 13(6), 385-396 (2013-05-25)
Understanding the biology of the breast and how ovarian hormones impinge on it is key to rational new approaches in breast cancer prevention and therapy. Because of the success of selective oestrogen receptor modulators (SERMs), such as tamoxifen, and aromatase
Marian Szamatowicz et al.
Ginekologia polska, 84(3), 219-222 (2013-05-25)
Uterine leiomyomata (fibroids) are very common, mostly benign tumors in women of reproductive age. Symptomatic fibroids cause significant morbidity and are characterized by heavy prolonged menstrual bleeding, by pain and pelvic pressure and, in some cases, they may lead to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service