Skip to Content
Merck
All Photos(1)

Documents

O-039

Supelco

Oxcarbazepine-13C6 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

¹³C₆ Oxcarbazepine Solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C9H12N2O2
Molecular Weight:
258.22
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

This stable-labeled internal standard is suitable for use in quantitation of oxcarbazepine levels or isotope dilution methods by GC/MS or LC/MS for applications in pain prescription monitoring, clinical toxicology, therapeutic drug monitoring or urine drug testing. Oxcarbazepine, marketed under the trade name Trileptal®, is an anticonvulsant developed and prescribed for treatment of epilepsy. In recent years, Oxcarbazepine has shown efficacy in treatment of mood disorders.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Trileptal is a registered trademark of Novartis Corporation

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service