Skip to Content
Merck
All Photos(1)

Key Documents

C-121

Supelco

Carbamazepine-10,11-epoxide solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C(N)N1C2=CC=CC=C2C3C(O3)C4=CC=CC=C14

InChI

1S/C15H12N2O2/c16-15(18)17-11-7-3-1-5-9(11)13-14(19-13)10-6-2-4-8-12(10)17/h1-8,13-14H,(H2,16,18)

InChI key

ZRWWEEVEIOGMMT-UHFFFAOYSA-N

General description

Carbamazepine-10,11-epoxide is the primary, active metabolite in both urine and serum of carbamazepine, an anticonvulsant drug used in the treatment of epilepsy and bipolar disorder as well as trigeminal neuralgia. Potential toxicity of the epoxide metabolite requires regular therapeutic drug monitoring by LC or LC/MS for patients taking carbamazepine.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gwendolyn A McMillin et al.
Journal of clinical laboratory analysis, 25(3), 212-216 (2011-05-14)
Discordant carbamazepine values as determined by two different immunoassays may be due to different cross-reactivities with the active metabolite carbamazepine 10, 11-epoxide and may cause confusion in interpreting carbamazepine serum levels. In this study, we compared carbamazepine values in samples
Gwendolyn A McMillin et al.
American journal of clinical pathology, 133(5), 728-736 (2010-04-17)
Carbamazepine is metabolized to an active metabolite known as carbamazepine-10,11-epoxide, or simply the "epoxide" metabolite. The presence of this metabolite can have clinically significant implications in therapeutic drug monitoring of carbamazepine, but accurate quantification of the epoxide metabolite is currently
W Qiu et al.
Phytotherapy research : PTR, 23(11), 1553-1558 (2009-04-17)
The in vivo effects of berberine (BBR), the widely used bioactive herbal ingredient from many traditional Chinese medicinal herbs, on the pharmacokinetics of carbamazepine (CBZ, a substrate of CYP3A) and its metabolite carbamazepine 10,11-epoxide (ECBZ), digoxin (DIG, a substrate of
Sigrid Mennickent et al.
Journal of separation science, 32(9), 1454-1458 (2009-03-31)
An instrumental planar chromatographic (HPTLC) method for quantification of carbamazepine in human serum was developed using liquid-liquid extraction with dichloromethane, fluorescence activation with perchloric acid 60%/ethanol/water (1:1:1, v/v) and fluorescence detection. Planar chromatographic separation was performed on precoated silica gel
Ana Fortuna et al.
Analytical and bioanalytical chemistry, 397(4), 1605-1615 (2010-04-20)
For the first time, a simple, selective and accurate high-performance liquid chromatography method with ultraviolet detection was developed and validated to quantify simultaneously three structurally related antiepileptic drugs; carbamazepine, oxcarbazepine, and the recently launched eslicarbazepine acetate and their main metabolites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service