Skip to Content
Merck
All Photos(1)

Key Documents

N-038

Supelco

6β-Naltrexol solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25NO4
CAS Number:
Molecular Weight:
343.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

OC1=C(O2)C([C@]([C@@H]2[C@H](O)CC3)(CCN4CC5CC5)[C@]3(O)[C@H]4C6)=C6C=C1

InChI

1S/C20H25NO4/c22-13-4-3-12-9-15-20(24)6-5-14(23)18-19(20,16(12)17(13)25-18)7-8-21(15)10-11-1-2-11/h3-4,11,14-15,18,22-24H,1-2,5-10H2/t14-,15-,18+,19+,20-/m1/s1

InChI key

JLVNEHKORQFVQJ-PYIJOLGTSA-N

General description

6ß-Naltrexol is the primary urinary metabolite of Naltrexone, an opiate sold under the trade names Revia, Depade, and Vivitrol® and used primarily in the management of alcohol or opiate dependence. This analytical reference standard is appropriate for use in LC/MS or GC/MS applications from clinical toxicology and forensic analysis to urine drug testing.

Application



  • Advanced analytical techniques for pharmacokinetics: The development of a rapid and sensitive gas chromatography-mass spectrometry method for nalmefene quantification in human plasma, pivotal for pharmacokinetic studies, underscores the importance of 6β-Naltrexol in therapeutic drug monitoring and addiction treatment research (Xie et al., 2002).



Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vivitrol is a registered trademark of Alkermes, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul K Kiptoo et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 33(4-5), 371-379 (2008-03-07)
6-Beta-naltrexol is the major active metabolite of naltrexone, NTX, a potent mu-opioid receptor antagonist used in the treatment of alcohol dependence and opioid abuse. Compared to naloxone, NTX has a longer duration of action largely attributed to 6-beta-naltrexol. This study
Sonja Brünen et al.
Analytical and bioanalytical chemistry, 396(3), 1249-1257 (2009-12-01)
We present data for a comparison of a liquid-chromatographic method coupled with tandem mass spectrometry (LC-MS/MS) and a high-performance liquid-chromatographic method with column switching and UV spectrophotometric detection. The two methods were developed for determination of naltrexone and 6beta-naltrexol in
Andrea L Pelotte et al.
Bioorganic & medicinal chemistry letters, 19(10), 2811-2814 (2009-04-15)
Since the mu opioid receptor (MOR) is known to be involved in the therapeutically relevant pathways leading to the manifestation of pain and addiction, we are currently studying the specific structural characteristics that promote antagonism at the MOR. The opiates
Jennifer A Bayron et al.
Journal of chemical information and modeling, 52(2), 391-395 (2012-01-24)
Naltrexol and its C₆ α and β desoxy, iodo, mesyl, tosyl, trifyl, dimethylcarbamyl, and diphenylcarbamyl derivatives were studied in their energy-minimized C ring chair-like and boat-like conformations using B3LYP/6-31G** and SM5.4/A to estimate aqueous solvation free energy. The results were
Matthew H Slawson et al.
Journal of analytical toxicology, 31(8), 453-461 (2007-11-09)
To improve the analysis of naltrexone and its primary metabolite 6beta-naltrexol, a sensitive and specific method for the analysis of subnanogram-per-milliliter concentrations of these analytes in human, rat, and rabbit plasma was developed utilizing liquid chromatography (LC) coupled to electrospray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service