Skip to Content
Merck
All Photos(1)

Key Documents

N-005

Supelco

Norcodeine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

COc1ccc2C[C@H]3NCC[C@@]45[C@@H](Oc1c24)[C@@H](O)C=C[C@@H]35

InChI

1S/C17H19NO3/c1-20-13-5-2-9-8-11-10-3-4-12(19)16-17(10,6-7-18-11)14(9)15(13)21-16/h2-5,10-12,16,18-19H,6-8H2,1H3/t10-,11+,12-,16-,17-/m0/s1

InChI key

HKOIXWVRNLGFOR-KOFBORESSA-N

General description

Norcodeine is a primary urinary metabolite of codeine. Codeine is an opiate used for its analgesic, antitussive, and sedative properties and is the second-most predominant alkaloid in opium. This certified Snap-N-Spike® Solution is applicable for use in LC/MS or GC/MS applications from urine drug testing and pain prescription monitoring to clinical toxicology and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Morphinan alkaloids present in mammal].
Y M Ning et al.
Sheng li ke xue jin zhan [Progress in physiology], 21(4), 329-332 (1990-10-01)
J O Svensson et al.
Journal of chromatography. B, Biomedical applications, 674(1), 49-55 (1995-12-01)
A reversed-phase ion-pair high-performance liquid chromatographic method for the simultaneous determination of codeine and seven metabolites is described. The samples are purified by reversed-phase solid-phase extraction. Codeine, norcodeine, codeine-6-glucuronide, norcodeine-6-glucuronide and morphine-3-glucuronide are measured with UV detection. Detection limits are
Z R Chen et al.
Journal of chromatography, 491(2), 367-378 (1989-07-21)
A novel high-performance liquid chromatographic method for the determination of codeine, norcodeine and morphine in plasma and urine has been developed. The compounds were separated on a cyano column (15 cm x 4.6 mm, 5 microns particle size) using a
Y Caraco et al.
Drug metabolism and disposition: the biological fate of chemicals, 24(7), 761-764 (1996-07-01)
Codeine is metabolized by glucuronidation, by O-demethylation to morphine, and by N-demethylation to norcodeine. The enzyme responsible for the O-demethylation to morphine has been identified as cytochrome P4502D6 (CYP2D6). The purpose of the present study was to identify the specific
Determination of codeine and its metabolites in human blood plasma and in microsomal incubates by high-performance liquid chromatography with ultraviolet detection.
K Persson et al.
Journal of chromatography, 491(2), 473-480 (1989-07-21)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service