Skip to Content
Merck
All Photos(1)

Key Documents

N-059

Supelco

Norbuprenorphine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H35NO4
CAS Number:
Molecular Weight:
413.55
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

CO[C@]12CC[C@@]3(C[C@@H]1[C@](C)(O)C(C)(C)C)[C@H]4Cc5ccc(O)c6O[C@@H]2[C@]3(CCN4)c56

InChI

1S/C25H35NO4/c1-21(2,3)22(4,28)16-13-23-8-9-25(16,29-5)20-24(23)10-11-26-17(23)12-14-6-7-15(27)19(30-20)18(14)24/h6-7,16-17,20,26-28H,8-13H2,1-5H3/t16-,17-,20-,22+,23-,24+,25-/m1/s1

InChI key

YOYLLRBMGQRFTN-IOMBULRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A new Certified Spiking Solution® suitable for use in clinical toxicology, forensic testing, or pain prescription monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gwendolyn A McMillin et al.
Journal of analytical toxicology, 36(2), 81-87 (2012-02-18)
Patterns of buprenorphine and metabolites were examined in 1946 positive urine samples analyzed by liquid chromatography-tandem mass spectrometry for free (unconjugated) buprenorphine and norbuprenorphine (quantitative, 2 to 1000 ng/mL) and buprenorphine-glucuronide (B3G) and norbuprenorphine-glucuronide (N3G) (semi-quantitative, 5 to 1000 ng/mL).
Kristen M Messenger et al.
Veterinary anaesthesia and analgesia, 38(4), 374-384 (2011-04-20)
To describe the pharmacokinetics and adverse effects of intravenous (IV) and sublingual (SL) buprenorphine in horses, and to determine the effect of sampling site on plasma concentrations after SL administration. Randomized crossover experiment; prospective study. Eleven healthy adult horses between
Hong K Kim et al.
Pediatrics, 130(6), e1700-e1703 (2012-11-07)
Buprenorphine is a partial μ-opioid receptor agonist that is approved for the treatment of opioid dependency. It is generally believed to be safer than methadone because of its ceiling effect on respiratory depression. As more adults in US households use
Tor Seldén et al.
Forensic science international, 220(1-3), 284-290 (2012-05-09)
Buprenorphine is considered to have little respiratory side effects at therapeutic doses and the partial agonistic properties should produce a "ceiling effect" for respiratory depression at higher doses. Still, there are several reports on buprenorphine related deaths. Most deaths involve
Kenneth F Ilett et al.
Breastfeeding medicine : the official journal of the Academy of Breastfeeding Medicine, 7, 269-274 (2011-10-21)
The aim of the present study was to estimate the dose of buprenorphine and its primary metabolite norbuprenorphine that a breastfed infant would receive during maternal maintenance treatment with buprenorphine. Seven pregnant opioid-dependent women taking buprenorphine (median, 7 mg/day; range, 2.4-24 mg)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service