Skip to Content
Merck
All Photos(1)

Key Documents

M-199

Supelco

4-Methyl-N-ethyl-pentedrone (4-MEAP HCl) hydrochloride solution

1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21NO · HCl
CAS Number:
Molecular Weight:
255.78
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.N(C(CCC)C(=O)c1ccc(cc1)C)CC

InChI

1S/C14H21NO.ClH/c1-4-6-13(15-5-2)14(16)12-9-7-11(3)8-10-12;/h7-10,13,15H,4-6H2,1-3H3;1H

InChI key

ULTAUAJCWQFPHK-UHFFFAOYSA-N

General description

4-Methyl-N-ethyl-pentedrone, also known as 4-MEAP and 4-MEAPP, is a stimulant drug of the cathinone class and has emerged on the illicit drug market as a designer drug. 4-MEAP has been found to be incorrectly sold as 4-methylpentedrone. This Certified Spiking Solution; is suitable as starting material in calibrators and controls for GC/MS or LC/MS testing applications in forensic analysis, urine drug testing or clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camila C Figueiredo et al.
Microbiology (Reading, England), 153(Pt 9), 2910-2921 (2007-09-05)
Sporotrichosis, a mycosis caused by Sporothrix schenckii, is characterized by lymphocutaneous lesions. In immunocompromised hosts, this fungus may invade the bloodstream and disseminate to other tissues, such as lung and bone. Our group previously showed that S. schenckii yeasts adhere
C Fiset et al.
The Journal of physiology, 500 ( Pt 1), 51-64 (1997-04-01)
1. The hypothesis that Kv4.2 and Kv4.3 are two of the essential K+ channel isoforms underlying the Ca2+-independent transient outward K+ current (It) in rat ventricle has been tested using a combination of electrophysiological measurements and antisense technology in both
Piotr Kaczynski et al.
International journal of molecular sciences, 21(3) (2020-02-06)
Estradiol-17β (E2) is a key hormone regulating reproductive functions in females. In pigs, E2, as the main conceptus signal, initiates processes resulting in prolonged corpus luteum function, embryo development, and implantation. During early pregnancy the endometrium undergoes morphological and physiological
Soledad Henríquez et al.
Human reproduction (Oxford, England), 35(7), 1655-1665 (2020-06-20)
Do alterations in pro- and anti-angiogenic estrogen metabolites in follicular fluid (FF) contribute to the follicular growth arrest and anovulation associated with polycystic ovary syndrome (PCOS)? FF of PCOS women with anovulation have reduced levels of pro-angiogenic estrogen metabolites (EMs)
M Lucas et al.
British journal of haematology, 110(4), 957-964 (2000-10-29)
The effect of overexpression of heat shock protein (HSP)72 on apoptosis induced by different stimuli in human umbilical vein endothelial cells (HUVECs) and the angiogenic cell line, ECV304, was studied. Transient overexpression of HSP72 was achieved using an adenoviral vector

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service