Skip to Content
Merck
All Photos(1)

Key Documents

D-081

Supelco

Dopamine hydrochloride solution

1.0 mg/mL in methanol with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO2·HCl
CAS Number:
Molecular Weight:
189.64
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol with 5% 1 M HCl (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

Cl.NCCc1ccc(O)c(O)c1

InChI

1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H

InChI key

CTENFNNZBMHDDG-UHFFFAOYSA-N

Gene Information

human ... ADRB1(153)

General description

Dopamine is a catecholamine which acts as a neurotransmitter for dopamine receptors and neurohormone for regulation of numerous brain functions. This certified solution and linearity standard is suitable for LC/MS applications in diagnostic testing, endocrinology, and clinical chemistry.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanneke E M den Ouden et al.
Neuron, 80(4), 1090-1100 (2013-11-26)
Serotonin and dopamine are speculated to subserve motivationally opponent functions, but this hypothesis has not been directly tested. We studied the role of these neurotransmitters in probabilistic reversal learning in nearly 700 individuals as a function of two polymorphisms in
Andrew S Hart et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(3), 698-704 (2014-01-17)
Making predictions about the rewards associated with environmental stimuli and updating those predictions through feedback is an essential aspect of adaptive behavior. Theorists have argued that dopamine encodes a reward prediction error (RPE) signal that is used in such a
Eric Set et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(26), 9615-9620 (2014-07-01)
Game theory describes strategic interactions where success of players' actions depends on those of coplayers. In humans, substantial progress has been made at the neural level in characterizing the dopaminergic and frontostriatal mechanisms mediating such behavior. Here we combined computational
Celia Herrera-Rincon et al.
NPJ Regenerative medicine, 5, 2-2 (2020-02-13)
Infections have numerous effects on the brain. However, possible roles of the brain in protecting against infection, and the developmental origin and role of brain signaling in immune response, are largely unknown. We exploited a unique Xenopus embryonic model to
Takeshi Inoue et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(17), 5927-5937 (2014-04-25)
LMTK3 belongs to the LMTK family of protein kinases that are predominantly expressed in the brain. Physiological functions of LMTK3 and other members of the LMTK family in the CNS remain unknown. In this study, we performed a battery of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service