Skip to Content
Merck
All Photos(1)

Documents

C-242

Supelco

Cannabichromevarin (CBCV) solution

1 mg/mL (Degassed Methanol ), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (Degassed Methanol )

format

single component solution

storage temp.

-10 to -25°C

InChI

1S/C19H26O2/c1-5-7-15-12-17(20)16-9-11-19(4,21-18(16)13-15)10-6-8-14(2)3/h8-9,11-13,20H,5-7,10H2,1-4H3

InChI key

AAXZFUQLLRMVOG-UHFFFAOYSA-N

General description

Cannabichromevarin (CBCV), also known as cannabivarichromene, is a phytocannabinoid found naturally in cannabis, and is an effective anticonvulsant and used to treat brain cancer and epilepsy. This certified Snap-N-Spike® solution is suitable as starting material in preparation of calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis, clinical toxicology, pharmaceutical research to cannabis testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service