Skip to Content
Merck
All Photos(1)

Key Documents

LM4103

Avanti

7α-hydroxycholesterol (D7)

Avanti Research - A Croda Brand LM4103, methanol solution

Synonym(s):

cholest-5-en-3β,7α-diol(d7)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39D7O2
CAS Number:
Molecular Weight:
409.70
UNSPSC Code:
12352100
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM4103-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM4103

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=C[C@H]2O)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

General description

7α-hydroxycholesterol is an oxysterol synthesized by the oxidation of the side chain of cholesterol. It is the precursor of bile acid.

Application

7α-hydroxycholesterol (D7) may be used:
  • as a deuterated sterol standard for the extraction of lipids from cultured cells
  • in liquid chromatography-high-resolution mass spectrometry method (LC-MS/HR-MS) for the quantification
  • to synthesize d4-7 -hydroxy-3-oxo-4-cholestenoic acid (d4-7-HOCA)

Packaging

2 mL Amber Glass Sealed Ampule (LM4103-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of the 15 major human bile acids and their precursor 7? -hydroxy-4-cholesten-3-one in serum by liquid chromatography--tandem mass spectrometry
Steiner C, et al.
Journal of Chromatography. B, Biomedical Applications, 878(28), 2870-2880 (2010)
Oxysterols in the pathogenesis of major chronic diseases
Poli G, et al.
Redox Biology, 1(1), 125-130 (2013)
7 ? -hydroxy-3-oxo-4-cholestenoic acid in cerebrospinal fluid reflects the integrity of the blood-brain barrier
Saeed A, et al.
Journal of Lipid Research, 55(2), 313-318 (2014)
Jeffrey G McDonald et al.
Methods in enzymology, 432, 145-170 (2007-10-24)
We describe the development of a high performance liquid chromatography mass spectrometry (HPLC-MS) method that allows the identification and quantitation of sterols in mammalian cells and tissues. Bulk lipids are extracted from biological samples by a modified Bligh/Dyer procedure in
Zoltán Pataj et al.
Journal of chromatography. A, 1439, 82-88 (2015-11-27)
Oxysterols are important intermediates in numerous metabolic and catabolic pathways and their biological significance is also proven. The present paper describes a reliable and short liquid chromatography-high-resolution mass spectrometry method (LC-MS/HR-MS) for the quantification of 8 different oxysterols (24(S)-hydroxycholesterol, 25-hydroxycholesterol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service