Skip to Content
Merck
All Photos(1)

Key Documents

LM1102

Avanti

17:0-20:4 PE

Avanti Research - A Croda Brand LM1102, methanol solution

Synonym(s):

1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H76NO8P
CAS Number:
Molecular Weight:
754.03
UNSPSC Code:
12352211
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1102-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1102

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O

InChI

1S/C42H76NO8P/c1-3-5-7-9-11-13-15-17-19-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43)38-48-41(44)34-32-30-28-26-24-22-18-16-14-12-10-8-6-4-2/h11,13,17,19,21,23,27,29,40H,3-10,12,14-16,18,20,22,24-26,28,30-39,43H2,1-2H3,(H,46,47)/b13-1

InChI key

PWFGSGJBCRORHV-DVHMRFIGSA-N

Application

17:0-20:4 PE or 1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phosphoethanolamine has been used as an internal standard:
  • for phospholipid extraction from mitochondria for quantitative mass spectrometry (MS) analysis
  • for lipid analysis of murine brain lipid extract using Fourier transform-ion cyclotron resonance-mass spectrometry (FT-ICR-MS)
  • for lipid quantification using liquid chromatography- mass spectrometry (LC-MS)
  • in the preparation of total lipid extracts by LC-MS
  • for ratiometric quantification of phosphoethanolamine (PE) using LC-MS

Biochem/physiol Actions

Phosphoethanolamine (PE) acts as a substrate for several phospholipids of the cell membrane. It is associated with the phospholipid turnover. PE is also found to prevent tumor progression.

Packaging

2 mL Amber Glass Sealed Ampule (LM1102-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Di Ding et al.
Molecular neurobiology, 56(4), 2618-2639 (2018-07-28)
Neurons from the adult central nervous system (CNS) demonstrate limited mRNA transport and localized protein synthesis versus developing neurons, correlating with lower regenerative capacity. We found that deimination (posttranslational conversion of protein-bound arginine into citrulline) undergoes upregulation during early neuronal
Andreas Ullen et al.
Free radical biology & medicine, 49(11), 1655-1665 (2010-09-03)
Plasmalogens, 1-O-alk-1'-enyl-2-acyl-sn-glycerophospholipids, are significant constituents of cellular membranes and are essential for normal brain development. Plasmalogens, which contain a vinyl ether bond at the sn-1 position, are preferential targets for hypochlorous acid (HOCl), generated by myeloperoxidase (MPO) from H(2)O(2) and
Kallol Gupta et al.
Nature protocols, 13(5), 1106-1120 (2018-04-28)
With the recent success in determining membrane protein structures, further detailed understanding of the identity and function of the bound lipidome is essential. Using an approach that combines high-energy native mass spectrometry (HE-nMS) and solution-phase lipid profiling, this protocol can
Laura Montani et al.
The Journal of cell biology, 217(4), 1353-1368 (2018-02-13)
Myelination calls for a remarkable surge in cell metabolism to facilitate lipid and membrane production. Endogenous fatty acid (FA) synthesis represents a potentially critical process in myelinating glia. Using genetically modified mice, we show that Schwann cell (SC) intrinsic activity
Adilson Kleber Ferreira et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 67(6), 481-487 (2013-06-19)
Phosphoethanolamine (Pho-s) is a compound involved in phospholipid turnover, acting as a substrate for many phospholipids of the cell membranes. In a recent study, we showed that Pho-s has antitumor effect in the several tumor cells. In this study we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service