Merck
  • Synthetic phosphoethanolamine induces cell cycle arrest and apoptosis in human breast cancer MCF-7 cells through the mitochondrial pathway.
Synthetic phosphoethanolamine induces cell cycle arrest and apoptosis in human breast cancer MCF-7 cells through the mitochondrial pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2013-06-19)
Adilson Kleber Ferreira, Renato Meneguelo, Alexandre Pereira, Otaviano Mendonça R Filho, Gilberto Orivaldo Chierice, Durvanei Augusto Maria
ABSTRACT
Phosphoethanolamine (Pho-s) is a compound involved in phospholipid turnover, acting as a substrate for many phospholipids of the cell membranes. In a recent study, we showed that Pho-s has antitumor effect in the several tumor cells. In this study we evaluated the antitumor activity of synthetic Pho-s on MCF-7 breast cancer cells. Here we demonstrate that Pho-s is cytotoxic to MCF-7 cells in a dose-dependent manner, while it is cytotoxic to MCF10 only at higher concentrations. In addition, Pho-s induces a disruption in mitochondrial membrane potential (Δψm). Furthermore, Pho-s induces mitochondria aggregates in the cytoplasm and DNA fragmentation of MCF-7 cells visualized by confocal microscopy. In agreement with the reduction on Δψm, we showed that Pho-s induces apoptosis followed by an increase in cytochrome c expression and capase-3-like activity in MCF-7 cells. Our results demonstrate that Pho-s induces a cell cycle arrest in the G1 phase through an inhibition of cyclin D1 and stimulates p53. An additional highlight of this study is the finding that Pho-s inhibits Bcl-2, inducing apoptosis through the mitochondrial pathway. Taken together, these results show that Pho-s is a promising compound in the fight against cancer.
Product Description
Avanti
16:0-06:0 NBD PE, Avanti Research - A Croda Brand 810153C
Avanti
16:0-06:0 NBD PE, Avanti Research - A Croda Brand 810153P, powder
Avanti
16:0 PEG1000 PE, Avanti Research - A Croda Brand 880700P, powder
Avanti
18:1-06:0 NBD PE, Avanti Research - A Croda Brand 810155C
Avanti
18:0 PEG1000 PE, Avanti Research - A Croda Brand 880720P, powder
Avanti
18:1-12:0 NBD PE, Avanti Research - A Croda Brand 810156P, powder
Avanti
16:0-18:1 Diether PE, Avanti Research - A Croda Brand 999974P, powder
Avanti
17:0-20:4 PE, Avanti Research - A Croda Brand LM1102, methanol solution