Skip to Content
Merck
All Photos(2)

Key Documents

890000P

Avanti

MVL5

Avanti Research - A Croda Brand 890000P, powder

Synonym(s):

N1-[2-((1S)-1-[(3-aminopropyl)amino]-4-[di(3-amino-propyl)amino]butylcarboxamido)ethyl]-3,4-di[oleyloxy]-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H116Cl5N7O4
CAS Number:
Molecular Weight:
1164.86
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

package of 1 × 1 mg (890000P-1mg)
package of 1 × 5 mg (890000P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand 890000P

application(s)

advanced drug delivery

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCC/C=C\CCCCCCCCOC1=C(OCCCCCCCC/C=C\CCCCCCCC)C=C(C(NCCNC(C([NH2+]CCC[NH3+])CCC[N+](CCC[NH3+])([H])CCC[NH3+])=O)=O)C=C1.[5Cl-]

Application

MVL5 has been used in liposome preparation.

Biochem/physiol Actions

MVL5 is a cationic lipid and is a pentavalent molecule. It is combined with 1,2-dioleoyl-sn-glycero-3-ethylphosphocholine (DOPC) in liposome formulation, for efficient gene transfection. It is considered less toxic.

Packaging

5 mL Amber Glass Screw Cap Vial (890000P-1MG)
5 mL Amber Glass Screw Cap Vial (890000P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Ling Chan et al.
The journal of gene medicine, 16(3-4), 84-96 (2014-04-23)
Cationic liposome (CL)-DNA complexes are promising gene delivery vectors with potential application in gene therapy. A key challenge in creating CL-DNA complexes for application is that their transfection efficiency (TE) is adversely affected by serum. In particular, little is known
Polyamine Drug Discovery, 226-226 (2012)
Asymmetric cationic liposomes designed for heat-activated association with cells
Jing Y, et al.
Colloids and Surfaces. B, Biointerfaces, 151, 112-118 (2017)
Nathan F Bouxsein et al.
Biochemistry, 46(16), 4785-4792 (2007-03-30)
Small interfering RNAs (siRNAs) of 19-25 bp mediate the cleavage of complementary mRNA, leading to post-transcriptional gene silencing. We examined cationic lipid (CL)-mediated delivery of siRNA into mammalian cells and made comparisons to CL-based DNA delivery. The effect of lipid
Endosomal escape and transfection efficiency of PEGylated cationic liposome-DNA complexes prepared with an acid-labile PEG-lipid
Chan CL, et al.
Biomaterials, 33(19), 4928-4935 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service