Skip to Content
Merck
All Photos(1)

Key Documents

870282P

Avanti

18:1 Biotinyl PE

Avanti Research - A Croda Brand 870282P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(biotinyl) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H91N3O10PNaS
CAS Number:
Molecular Weight:
992.31
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 100 mg (870282P-100mg)
pkg of 2 × 100 mg (870282P-200mg)
pkg of 1 × 25 mg (870282P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870282P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCC[C@](SC[C@@]1(NC(N2)=O)[H])([C@]12[H])[H])=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C51H92N3O10PS.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-37-48(56)61-41-44(64-49(57)38-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)42-63-65(59,60)62-40-39-52-47(55)36-34-33-35-46-50-45(43-66-46)53-51(58)54-50;/h17-20,44-46,50H,3-16,21-43H2,1-2H3,(H,52,55)(H,59,60)(H2,53,54,58);/q;+1/p-1/b19-17-,20-18-;/t44-,45+,46+,50+;/m1./s1

InChI key

ZGFSYDGJPSEMJM-AOAFAMOJSA-M

Application

18:1 Biotinyl PE may be used to tag N-glycolyl-monosialodihexosyl-ganglioside (NGcGM3) in the competitive assay.[1] It may also be used in the preparation of biotin-containing phospholipid vesicle layer for biosensing.[2]

Packaging

5 mL Clear Glass Sealed Ampule (870282P-100mg)
5 mL Clear Glass Sealed Ampule (870282P-200mg)
5 mL Clear Glass Sealed Ampule (870282P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Virginia Gentilini et al.
Cancer immunology, immunotherapy : CII, 65(5), 551-562 (2016-03-13)
The expression of N-glycolyl-monosialodihexosyl-ganglioside (NGcGM3) in humans is restricted to cancer cells; therefore, it is a tumor antigen. There are measurable quantities of circulating anti-NGcGM3 antibodies (aNGcGM3 Abs) in human serum. Interestingly, some people have circulating Ag-specific immunoglobulins G (IgGs)
Yoshiko Ishizuka-Katsura et al.
Journal of bioscience and bioengineering, 105(5), 527-535 (2008-06-19)
We describe a technique to form a biotin-containing phospholipid vesicle layer on a self-assembled monolayer (SAM) deposited on a gold surface to immobilize biotinylated receptor proteins for a surface plasmon resonance (SPR) biosensor. The adsorption of vesicle of 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC)
Ingrid Albertsson et al.
Scientific reports, 9(1), 17234-17234 (2019-11-23)
Denitrification is a microbial pathway that constitutes an important part of the nitrogen cycle on earth. Denitrifying organisms use nitrate as a terminal electron acceptor and reduce it stepwise to nitrogen gas, a process that produces the toxic nitric oxide
Heun Jin Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(49), 19253-19257 (2008-12-03)
Recent advances have enabled 3-dimensional reconstructions of biological structures in vivo, ranging in size and complexity from single proteins to multicellular structures. In particular, tomography and confocal microscopy have been exploited to capture detailed 3-dimensional conformations of membranes in cellular
Zachary I Imam et al.
Langmuir : the ACS journal of surfaces and colloids, 35(49), 16281-16289 (2019-11-16)
Cytoskeletal filaments and motor proteins are critical components in the transport and reorganization of membrane-based organelles in eukaryotic cells. Previous studies have recapitulated the microtubule-kinesin transport system in vitro to dynamically assemble large-scale nanotube networks from multilamellar liposomes and polymersomes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service